Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38837263-38837536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPL
Entrez Gene ID
3191
Gene Name
heterogeneous nuclear ribonucleoprotein L
Gene Aliases
HNRPL, P/OKcl.14, hnRNP-L
UniGene
Hs.644906
Gene Chromosome Location
Chr.19: 38836378-38852339 on Build GRCh38
Transcripts
NM_001005335.1 NM_001533.2 XM_011526886.1 XM_011526887.1 XM_011526888.2 XM_011526889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6151190 c.1282T>A p.F428I Chr.19: 38837414-38837414 on Build GRCh38
6151189 c.1681T>A p.F561I Chr.19: 38837414-38837414 on Build GRCh38
6681344 c.1219G>A p.E407K Chr.19: 38837477-38837477 on Build GRCh38
4926623 c.1675C>G p.L559V Chr.19: 38837420-38837420 on Build GRCh38
5755361 c.1701G>T p.M567I Chr.19: 38837394-38837394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777969085 Chr.19: 38837435-38837435 on Build GRCh38
rs765464827 Chr.19: 38837388-38837388 on Build GRCh38
rs762825760 Chr.19: 38837310-38837311 on Build GRCh38
rs755291333 Chr.19: 38837387-38837387 on Build GRCh38
rs531405956 Chr.19: 38837377-38837377 on Build GRCh38
rs759378514 Chr.19: 38837318-38837318 on Build GRCh38
rs751027234 Chr.19: 38837362-38837362 on Build GRCh38
rs990482433 Chr.19: 38837413-38837413 on Build GRCh38
rs764751159 Chr.19: 38837396-38837396 on Build GRCh38
rs775012093 Chr.19: 38837496-38837496 on Build GRCh38
rs748820618 Chr.19: 38837493-38837493 on Build GRCh38
rs1023324219 Chr.19: 38837420-38837420 on Build GRCh38
rs745680829 Chr.19: 38837475-38837475 on Build GRCh38
rs776621392 Chr.19: 38837310-38837310 on Build GRCh38
rs142449808 Chr.19: 38837427-38837427 on Build GRCh38
rs776223442 Chr.19: 38837286-38837286 on Build GRCh38
rs2278010 Chr.19: 38837505-38837505 on Build GRCh38
rs901383886 Chr.19: 38837323-38837323 on Build GRCh38
rs1005603345 Chr.19: 38837421-38837421 on Build GRCh38
rs997052909 Chr.19: 38837352-38837352 on Build GRCh38
rs764063333 Chr.19: 38837317-38837321 on Build GRCh38
rs775235387 Chr.19: 38837334-38837334 on Build GRCh38
rs761300230 Chr.19: 38837371-38837371 on Build GRCh38
rs766848522 Chr.19: 38837373-38837373 on Build GRCh38
rs747042478 Chr.19: 38837436-38837436 on Build GRCh38
rs981863850 Chr.19: 38837459-38837459 on Build GRCh38
rs759391418 Chr.19: 38837304-38837304 on Build GRCh38
rs781236253 Chr.19: 38837463-38837463 on Build GRCh38
rs970703166 Chr.19: 38837422-38837422 on Build GRCh38
rs372573886 Chr.19: 38837339-38837339 on Build GRCh38
rs903236520 Chr.19: 38837285-38837285 on Build GRCh38
rs769602768 Chr.19: 38837329-38837329 on Build GRCh38
rs752903039 Chr.19: 38837401-38837401 on Build GRCh38
rs151308132 Chr.19: 38837430-38837430 on Build GRCh38
rs761663334 Chr.19: 38837500-38837501 on Build GRCh38
rs749034719 Chr.19: 38837498-38837498 on Build GRCh38
rs751320266 Chr.19: 38837373-38837374 on Build GRCh38
rs952475137 Chr.19: 38837391-38837391 on Build GRCh38
rs777582947 Chr.19: 38837301-38837301 on Build GRCh38
rs763594494 Chr.19: 38837342-38837342 on Build GRCh38
rs757304417 Chr.19: 38837438-38837438 on Build GRCh38
rs1015279604 Chr.19: 38837425-38837425 on Build GRCh38

View Full Product Details