Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36855574-36855799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO1
Entrez Gene ID
9844
Gene Name
engulfment and cell motility 1
Gene Aliases
CED-12, CED12, ELMO-1
UniGene
Hs.434989
Gene Chromosome Location
Chr.7: 36852906-37449409 on Build GRCh38
Transcripts
NM_001039459.2 NM_001206480.2 NM_001206482.1 NM_014800.10 NM_130442.3 NR_038120.1 XM_005249919.2 XM_006715805.1 XM_011515654.1 XM_017012838.1 XM_017012839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6436826 c.2038G>A p.D680N Chr.7: 36855697-36855697 on Build GRCh38
7366129 c.1127G>T p.G376V Chr.7: 36855714-36855714 on Build GRCh38
6110339 c.1207C>A p.L403M Chr.7: 36855634-36855634 on Build GRCh38
5358159 c.2067G>A p.L689L Chr.7: 36855668-36855668 on Build GRCh38
4786996 c.1104G>A p.T368T Chr.7: 36855737-36855737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757503801 Chr.7: 36855661-36855661 on Build GRCh38
rs201616410 Chr.7: 36855687-36855687 on Build GRCh38
rs764481575 Chr.7: 36855730-36855730 on Build GRCh38
rs751754505 Chr.7: 36855645-36855645 on Build GRCh38
rs750500849 Chr.7: 36855757-36855757 on Build GRCh38
rs765898655 Chr.7: 36855619-36855619 on Build GRCh38
rs137942550 Chr.7: 36855683-36855683 on Build GRCh38
rs776112181 Chr.7: 36855722-36855722 on Build GRCh38
rs749009029 Chr.7: 36855692-36855692 on Build GRCh38
rs757295660 Chr.7: 36855723-36855723 on Build GRCh38
rs986799793 Chr.7: 36855670-36855670 on Build GRCh38
rs767784847 Chr.7: 36855738-36855738 on Build GRCh38
rs756037174 Chr.7: 36855689-36855689 on Build GRCh38
rs984811545 Chr.7: 36855739-36855739 on Build GRCh38
rs149257728 Chr.7: 36855611-36855611 on Build GRCh38
rs752998607 Chr.7: 36855620-36855620 on Build GRCh38
rs866035568 Chr.7: 36855704-36855704 on Build GRCh38
rs1054649854 Chr.7: 36855701-36855701 on Build GRCh38
rs13246439 Chr.7: 36855737-36855737 on Build GRCh38
rs911454272 Chr.7: 36855671-36855671 on Build GRCh38
rs770393657 Chr.7: 36855721-36855721 on Build GRCh38
rs374523052 Chr.7: 36855774-36855774 on Build GRCh38
rs918674508 Chr.7: 36855610-36855610 on Build GRCh38
rs768548471 Chr.7: 36855697-36855697 on Build GRCh38
rs772830948 Chr.7: 36855716-36855716 on Build GRCh38
rs774585018 Chr.7: 36855734-36855734 on Build GRCh38
rs964094394 Chr.7: 36855608-36855608 on Build GRCh38
rs142425368 Chr.7: 36855719-36855719 on Build GRCh38
rs182459066 Chr.7: 36855690-36855690 on Build GRCh38
rs892851067 Chr.7: 36855599-36855599 on Build GRCh38
rs747766842 Chr.7: 36855710-36855710 on Build GRCh38
rs778602057 Chr.7: 36855698-36855698 on Build GRCh38
rs377709624 Chr.7: 36855602-36855602 on Build GRCh38
rs771861381 Chr.7: 36855712-36855712 on Build GRCh38
rs34022626 Chr.7: 36855669-36855670 on Build GRCh38
rs143442077 Chr.7: 36855621-36855621 on Build GRCh38
rs371172929 Chr.7: 36855674-36855674 on Build GRCh38

View Full Product Details