Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15649732-15649999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F3
Entrez Gene ID
4051
Gene Name
cytochrome P450 family 4 subfamily F member 3
Gene Aliases
CPF3, CYP4F, LTB4H
UniGene
Hs.106242
Gene Chromosome Location
Chr.19: 15640897-15660760 on Build GRCh38
Transcripts
NM_000896.2 NM_001199208.1 NM_001199209.1 XM_005259911.4 XM_011528014.2 XM_017026814.1 XM_017026815.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4075080 c.694G>A p.V232M Chr.19: 15649959-15649959 on Build GRCh38
7144908 c.684C>T p.L228L Chr.19: 15649949-15649949 on Build GRCh38
3772528 c.686G>A p.S229N Chr.19: 15649951-15649951 on Build GRCh38
3772529 c.686G>A p.S229N Chr.19: 15649951-15649951 on Build GRCh38
6277597 c.651G>A p.K217K Chr.19: 15649916-15649916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904417237 Chr.19: 15649927-15649927 on Build GRCh38
rs746700434 Chr.19: 15649958-15649958 on Build GRCh38
rs1026670280 Chr.19: 15649827-15649827 on Build GRCh38
rs1040809786 Chr.19: 15649816-15649816 on Build GRCh38
rs973809218 Chr.19: 15649826-15649826 on Build GRCh38
rs771684813 Chr.19: 15649876-15649876 on Build GRCh38
rs760041800 Chr.19: 15649881-15649881 on Build GRCh38
rs866252258 Chr.19: 15649941-15649941 on Build GRCh38
rs772761668 Chr.19: 15649880-15649880 on Build GRCh38
rs928453780 Chr.19: 15649920-15649920 on Build GRCh38
rs939816463 Chr.19: 15649792-15649792 on Build GRCh38
rs960994407 Chr.19: 15649814-15649814 on Build GRCh38
rs979990914 Chr.19: 15649879-15649879 on Build GRCh38
rs747228958 Chr.19: 15649938-15649939 on Build GRCh38
rs202115280 Chr.19: 15649952-15649952 on Build GRCh38
rs1028895696 Chr.19: 15649757-15649757 on Build GRCh38
rs903966550 Chr.19: 15649974-15649974 on Build GRCh38
rs751811670 Chr.19: 15649931-15649931 on Build GRCh38
rs777799966 Chr.19: 15649926-15649926 on Build GRCh38
rs757242505 Chr.19: 15649933-15649933 on Build GRCh38
rs758681598 Chr.19: 15649925-15649925 on Build GRCh38
rs769018138 Chr.19: 15649959-15649959 on Build GRCh38
rs140136303 Chr.19: 15649946-15649946 on Build GRCh38
rs769074076 Chr.19: 15649957-15649957 on Build GRCh38
rs199836042 Chr.19: 15649873-15649873 on Build GRCh38
rs527733036 Chr.19: 15649953-15649953 on Build GRCh38
rs759544232 Chr.19: 15649962-15649962 on Build GRCh38
rs1008893978 Chr.19: 15649769-15649769 on Build GRCh38
rs144295345 Chr.19: 15649813-15649813 on Build GRCh38
rs776323965 Chr.19: 15649961-15649961 on Build GRCh38
rs10401516 Chr.19: 15649767-15649767 on Build GRCh38
rs371729462 Chr.19: 15649934-15649934 on Build GRCh38
rs368167752 Chr.19: 15649922-15649922 on Build GRCh38
rs776041725 Chr.19: 15649935-15649935 on Build GRCh38
rs200161271 Chr.19: 15649916-15649916 on Build GRCh38
rs551654869 Chr.19: 15649912-15649912 on Build GRCh38
rs765949141 Chr.19: 15649907-15649907 on Build GRCh38
rs759597693 Chr.19: 15649914-15649914 on Build GRCh38
rs969763143 Chr.19: 15649871-15649871 on Build GRCh38
rs908686278 Chr.19: 15649779-15649779 on Build GRCh38
rs1015263018 Chr.19: 15649774-15649774 on Build GRCh38

View Full Product Details