Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90539972-90540233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC3
Entrez Gene ID
64784
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
TORC-3, TORC3
UniGene
Hs.567572
Gene Chromosome Location
Chr.15: 90529886-90645345 on Build GRCh38
Transcripts
NM_001042574.2 NM_022769.4 XM_005254968.2 XM_011521906.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4827304 c.174C>T p.Y58Y Chr.15: 90540080-90540080 on Build GRCh38
6843094 c.160C>A p.R54S Chr.15: 90540066-90540066 on Build GRCh38
1629742 c.173A>G p.Y58C Chr.15: 90540079-90540079 on Build GRCh38
3999686 c.215G>A p.S72N Chr.15: 90540121-90540121 on Build GRCh38
6900736 c.187T>C p.L63L Chr.15: 90540093-90540093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759378312 Chr.15: 90540070-90540070 on Build GRCh38
rs1019231771 Chr.15: 90540194-90540194 on Build GRCh38
rs999379248 Chr.15: 90540201-90540201 on Build GRCh38
rs776504911 Chr.15: 90540067-90540067 on Build GRCh38
rs778167567 Chr.15: 90540178-90540179 on Build GRCh38
rs368636986 Chr.15: 90540090-90540090 on Build GRCh38
rs902216414 Chr.15: 90540199-90540199 on Build GRCh38
rs192214530 Chr.15: 90540125-90540125 on Build GRCh38
rs769643937 Chr.15: 90540032-90540032 on Build GRCh38
rs1050209961 Chr.15: 90540065-90540065 on Build GRCh38
rs748252016 Chr.15: 90540162-90540162 on Build GRCh38
rs774281439 Chr.15: 90540021-90540021 on Build GRCh38
rs142274876 Chr.15: 90540082-90540082 on Build GRCh38
rs376919825 Chr.15: 90540019-90540019 on Build GRCh38
rs374447890 Chr.15: 90540078-90540078 on Build GRCh38
rs540217467 Chr.15: 90540014-90540014 on Build GRCh38
rs151211329 Chr.15: 90540083-90540083 on Build GRCh38
rs749581771 Chr.15: 90540115-90540115 on Build GRCh38
rs898647383 Chr.15: 90540063-90540063 on Build GRCh38
rs780277574 Chr.15: 90540114-90540114 on Build GRCh38
rs761886760 Chr.15: 90540047-90540047 on Build GRCh38
rs764988454 Chr.15: 90540074-90540074 on Build GRCh38
rs766335111 Chr.15: 90540061-90540061 on Build GRCh38
rs779155715 Chr.15: 90540146-90540146 on Build GRCh38
rs565391111 Chr.15: 90540015-90540015 on Build GRCh38
rs750942610 Chr.15: 90540102-90540102 on Build GRCh38
rs8033595 Chr.15: 90540121-90540121 on Build GRCh38
rs1013530227 Chr.15: 90540200-90540200 on Build GRCh38
rs931451546 Chr.15: 90540066-90540066 on Build GRCh38
rs532538850 Chr.15: 90540076-90540076 on Build GRCh38
rs763741461 Chr.15: 90540079-90540079 on Build GRCh38
rs142472120 Chr.15: 90540110-90540110 on Build GRCh38
rs772117713 Chr.15: 90540179-90540179 on Build GRCh38
rs374582569 Chr.15: 90540080-90540080 on Build GRCh38
rs778318134 Chr.15: 90540035-90540035 on Build GRCh38

View Full Product Details