Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7283194-7283459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A4
Entrez Gene ID
6517
Gene Name
solute carrier family 2 member 4
Gene Aliases
GLUT4
UniGene
Hs.380691
Gene Chromosome Location
Chr.17: 7281735-7288048 on Build GRCh38
Transcripts
NM_001042.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4435428 c.34-3C>T p.? Chr.17: 7283242-7283242 on Build GRCh38
4069252 c.34-1G>T p.? Chr.17: 7283244-7283244 on Build GRCh38
349429 c.115G>T p.G39W Chr.17: 7283326-7283326 on Build GRCh38
3521666 c.104C>T p.S35F Chr.17: 7283315-7283315 on Build GRCh38
4451717 c.136A>T p.N46Y Chr.17: 7283347-7283347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991276496 Chr.17: 7283418-7283418 on Build GRCh38
rs755074748 Chr.17: 7283271-7283271 on Build GRCh38
rs1012726814 Chr.17: 7283420-7283420 on Build GRCh38
rs767051481 Chr.17: 7283219-7283219 on Build GRCh38
rs775018856 Chr.17: 7283304-7283304 on Build GRCh38
rs1041707570 Chr.17: 7283347-7283347 on Build GRCh38
rs756582644 Chr.17: 7283429-7283429 on Build GRCh38
rs13306762 Chr.17: 7283422-7283422 on Build GRCh38
rs767147777 Chr.17: 7283231-7283231 on Build GRCh38
rs763807880 Chr.17: 7283254-7283254 on Build GRCh38
rs750395028 Chr.17: 7283348-7283348 on Build GRCh38
rs757179263 Chr.17: 7283255-7283255 on Build GRCh38
rs147146068 Chr.17: 7283343-7283343 on Build GRCh38
rs751969732 Chr.17: 7283385-7283386 on Build GRCh38
rs139011011 Chr.17: 7283266-7283266 on Build GRCh38
rs374096878 Chr.17: 7283400-7283400 on Build GRCh38
rs760021028 Chr.17: 7283392-7283393 on Build GRCh38
rs768116687 Chr.17: 7283400-7283402 on Build GRCh38
rs921046882 Chr.17: 7283273-7283273 on Build GRCh38
rs371315465 Chr.17: 7283434-7283434 on Build GRCh38
rs1016481590 Chr.17: 7283356-7283356 on Build GRCh38
rs376636615 Chr.17: 7283315-7283315 on Build GRCh38
rs776290317 Chr.17: 7283437-7283437 on Build GRCh38
rs373915708 Chr.17: 7283435-7283435 on Build GRCh38
rs903206032 Chr.17: 7283275-7283275 on Build GRCh38
rs947358493 Chr.17: 7283262-7283262 on Build GRCh38
rs376727989 Chr.17: 7283427-7283427 on Build GRCh38
rs201878149 Chr.17: 7283225-7283225 on Build GRCh38
rs373585588 Chr.17: 7283411-7283411 on Build GRCh38
rs771486330 Chr.17: 7283317-7283317 on Build GRCh38
rs760269825 Chr.17: 7283433-7283433 on Build GRCh38
rs765115135 Chr.17: 7283346-7283346 on Build GRCh38
rs773966408 Chr.17: 7283223-7283223 on Build GRCh38
rs781146169 Chr.17: 7283385-7283385 on Build GRCh38
rs765043342 Chr.17: 7283256-7283259 on Build GRCh38
rs753595456 Chr.17: 7283340-7283340 on Build GRCh38
rs142195514 Chr.17: 7283277-7283277 on Build GRCh38
rs200125590 Chr.17: 7283438-7283438 on Build GRCh38
rs375712217 Chr.17: 7283407-7283407 on Build GRCh38
rs533993099 Chr.17: 7283336-7283336 on Build GRCh38
rs745856223 Chr.17: 7283417-7283417 on Build GRCh38
rs192326501 Chr.17: 7283283-7283283 on Build GRCh38
rs778692114 Chr.17: 7283413-7283413 on Build GRCh38
rs367888501 Chr.17: 7283372-7283372 on Build GRCh38
rs753201039 Chr.17: 7283422-7283423 on Build GRCh38
rs781438718 Chr.17: 7283274-7283274 on Build GRCh38
rs752882369 Chr.17: 7283391-7283391 on Build GRCh38
rs749618851 Chr.17: 7283402-7283402 on Build GRCh38
rs562894482 Chr.17: 7283245-7283245 on Build GRCh38
rs779495228 Chr.17: 7283409-7283409 on Build GRCh38
rs755868694 Chr.17: 7283249-7283249 on Build GRCh38
rs756288065 Chr.17: 7283395-7283395 on Build GRCh38
rs775146734 Chr.17: 7283334-7283334 on Build GRCh38
rs372415018 Chr.17: 7283399-7283399 on Build GRCh38
rs774855149 Chr.17: 7283431-7283431 on Build GRCh38
rs371883816 Chr.17: 7283229-7283229 on Build GRCh38
rs770138000 Chr.17: 7283280-7283280 on Build GRCh38
rs778865343 Chr.17: 7283261-7283261 on Build GRCh38
rs541196033 Chr.17: 7283335-7283335 on Build GRCh38
rs1007734551 Chr.17: 7283351-7283351 on Build GRCh38
rs143506382 Chr.17: 7283284-7283284 on Build GRCh38

View Full Product Details