Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 80337534-80337759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRY2
Entrez Gene ID
10253
Gene Name
sprouty RTK signaling antagonist 2
Gene Aliases
IGAN3, hSPRY2
UniGene
Hs.18676
Gene Chromosome Location
Chr.13: 80335976-80341115 on Build GRCh38
Transcripts
NM_001318536.1 NM_001318537.1 NM_001318538.1 NM_005842.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4923208 c.172G>T p.G58W Chr.13: 80337534-80337534 on Build GRCh38
4771514 c.118G>T p.V40F Chr.13: 80337588-80337588 on Build GRCh38
7200013 c.92G>C p.R31T Chr.13: 80337614-80337614 on Build GRCh38
6521148 c.76G>T p.G26W Chr.13: 80337630-80337630 on Build GRCh38
3469869 c.32C>T p.S11L Chr.13: 80337674-80337674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200366516 Chr.13: 80337571-80337571 on Build GRCh38
rs763949772 Chr.13: 80337616-80337616 on Build GRCh38
rs757245505 Chr.13: 80337632-80337632 on Build GRCh38
rs369819185 Chr.13: 80337723-80337723 on Build GRCh38
rs567043511 Chr.13: 80337680-80337680 on Build GRCh38
rs9574632 Chr.13: 80337688-80337688 on Build GRCh38
rs771424788 Chr.13: 80337678-80337678 on Build GRCh38
rs141948380 Chr.13: 80337687-80337687 on Build GRCh38
rs372480996 Chr.13: 80337656-80337656 on Build GRCh38
rs767200640 Chr.13: 80337633-80337633 on Build GRCh38
rs141331436 Chr.13: 80337647-80337647 on Build GRCh38
rs759745233 Chr.13: 80337608-80337608 on Build GRCh38
rs775821610 Chr.13: 80337708-80337708 on Build GRCh38
rs776681361 Chr.13: 80337607-80337607 on Build GRCh38
rs771050581 Chr.13: 80337594-80337594 on Build GRCh38
rs772271382 Chr.13: 80337582-80337582 on Build GRCh38
rs564716686 Chr.13: 80337667-80337667 on Build GRCh38
rs759757857 Chr.13: 80337668-80337668 on Build GRCh38
rs1012981412 Chr.13: 80337660-80337660 on Build GRCh38
rs762829116 Chr.13: 80337615-80337615 on Build GRCh38
rs755783967 Chr.13: 80337561-80337561 on Build GRCh38
rs368770309 Chr.13: 80337650-80337650 on Build GRCh38
rs145173125 Chr.13: 80337673-80337673 on Build GRCh38
rs903065031 Chr.13: 80337592-80337592 on Build GRCh38
rs894564957 Chr.13: 80337651-80337651 on Build GRCh38
rs748556088 Chr.13: 80337572-80337572 on Build GRCh38
rs764037420 Chr.13: 80337718-80337718 on Build GRCh38
rs761632096 Chr.13: 80337733-80337733 on Build GRCh38
rs771376928 Chr.13: 80337610-80337610 on Build GRCh38
rs763062952 Chr.13: 80337711-80337711 on Build GRCh38
rs993786432 Chr.13: 80337677-80337677 on Build GRCh38
rs773743161 Chr.13: 80337586-80337586 on Build GRCh38
rs202021168 Chr.13: 80337692-80337692 on Build GRCh38
rs751596093 Chr.13: 80337621-80337621 on Build GRCh38
rs1040671271 Chr.13: 80337589-80337589 on Build GRCh38
rs376727332 Chr.13: 80337676-80337676 on Build GRCh38
rs774765586 Chr.13: 80337657-80337657 on Build GRCh38

View Full Product Details