Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66793678-66793919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf44-SGK3
Entrez Gene ID
100533105
Gene Name
C8orf44-SGK3 readthrough
Gene Aliases
-
UniGene
Hs.613417
Gene Chromosome Location
Chr.8: 66667552-66862022 on Build GRCh38
Transcripts
NM_001204173.1 NM_001033578.2 NM_013257.4 NM_170709.2

Gene Symbol
SGK3
Entrez Gene ID
23678
Gene Name
serum/glucocorticoid regulated kinase family member 3
Gene Aliases
CISK, SGK2, SGKL
UniGene
Hs.613417
Gene Chromosome Location
Chr.8: 66712418-66862022 on Build GRCh38
Transcripts
NM_001204173.1 NM_001033578.2 NM_013257.4 NM_170709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6031389 c.22G>A p.D8N Chr.8: 66793758-66793758 on Build GRCh38
7254678 c.26A>G p.Y9C Chr.8: 66793762-66793762 on Build GRCh38
6776747 c.5_6delAA p.D4fs*19 Chr.8: 66793741-66793742 on Build GRCh38
3925476 c.62C>T p.S21F Chr.8: 66793798-66793798 on Build GRCh38
4392583 c.19A>G p.M7V Chr.8: 66793755-66793755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766986095 Chr.8: 66793776-66793776 on Build GRCh38
rs776902799 Chr.8: 66793710-66793710 on Build GRCh38
rs770790969 Chr.8: 66793725-66793725 on Build GRCh38
rs979152693 Chr.8: 66793882-66793882 on Build GRCh38
rs146507110 Chr.8: 66793800-66793800 on Build GRCh38
rs771904845 Chr.8: 66793733-66793733 on Build GRCh38
rs756819800 Chr.8: 66793845-66793845 on Build GRCh38
rs374126174 Chr.8: 66793724-66793724 on Build GRCh38
rs772027552 Chr.8: 66793872-66793872 on Build GRCh38
rs747879410 Chr.8: 66793703-66793703 on Build GRCh38
rs771419726 Chr.8: 66793709-66793710 on Build GRCh38
rs777299349 Chr.8: 66793825-66793825 on Build GRCh38
rs746366957 Chr.8: 66793809-66793812 on Build GRCh38
rs377252014 Chr.8: 66793873-66793873 on Build GRCh38
rs373626095 Chr.8: 66793871-66793871 on Build GRCh38
rs774341369 Chr.8: 66793728-66793728 on Build GRCh38
rs369810824 Chr.8: 66793702-66793702 on Build GRCh38
rs765398043 Chr.8: 66793744-66793744 on Build GRCh38
rs138778635 Chr.8: 66793802-66793802 on Build GRCh38
rs752166025 Chr.8: 66793782-66793782 on Build GRCh38
rs553948504 Chr.8: 66793743-66793743 on Build GRCh38
rs770229184 Chr.8: 66793839-66793840 on Build GRCh38
rs759313083 Chr.8: 66793732-66793732 on Build GRCh38
rs759024297 Chr.8: 66793889-66793889 on Build GRCh38
rs777292409 Chr.8: 66793723-66793723 on Build GRCh38
rs769366639 Chr.8: 66793717-66793717 on Build GRCh38
rs142472352 Chr.8: 66793770-66793770 on Build GRCh38
rs755369708 Chr.8: 66793793-66793793 on Build GRCh38
rs1036217721 Chr.8: 66793780-66793780 on Build GRCh38
rs765422599 Chr.8: 66793753-66793753 on Build GRCh38
rs750459045 Chr.8: 66793764-66793764 on Build GRCh38
rs778799926 Chr.8: 66793848-66793848 on Build GRCh38
rs370247117 Chr.8: 66793837-66793837 on Build GRCh38
rs776685803 Chr.8: 66793741-66793741 on Build GRCh38
rs769990612 Chr.8: 66793881-66793881 on Build GRCh38
rs775028323 Chr.8: 66793860-66793860 on Build GRCh38
rs367954710 Chr.8: 66793811-66793811 on Build GRCh38
rs746791326 Chr.8: 66793875-66793875 on Build GRCh38

View Full Product Details