Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3965779-3966014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY9
Entrez Gene ID
115
Gene Name
adenylate cyclase 9
Gene Aliases
AC9, ACIX
UniGene
Hs.391860
Gene Chromosome Location
Chr.16: 3953387-4116185 on Build GRCh38
Transcripts
NM_001116.3 XM_005255079.3 XM_011522353.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2920104 c.4043A>G p.N1348S Chr.16: 3965794-3965794 on Build GRCh38
73590 c.3930A>T p.R1310S Chr.16: 3965907-3965907 on Build GRCh38
158933 c.3940G>A p.E1314K Chr.16: 3965897-3965897 on Build GRCh38
1740229 c.4028A>G p.E1343G Chr.16: 3965809-3965809 on Build GRCh38
1629893 c.3830A>T p.E1277V Chr.16: 3966007-3966007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759474569 Chr.16: 3965875-3965875 on Build GRCh38
rs777093117 Chr.16: 3965928-3965928 on Build GRCh38
rs752822315 Chr.16: 3965803-3965803 on Build GRCh38
rs185283666 Chr.16: 3965817-3965817 on Build GRCh38
rs756290470 Chr.16: 3965930-3965930 on Build GRCh38
rs146352467 Chr.16: 3965882-3965882 on Build GRCh38
rs888717225 Chr.16: 3965948-3965948 on Build GRCh38
rs766303375 Chr.16: 3965897-3965897 on Build GRCh38
rs375515956 Chr.16: 3965993-3965993 on Build GRCh38
rs774279006 Chr.16: 3965872-3965872 on Build GRCh38
rs915968908 Chr.16: 3965894-3965894 on Build GRCh38
rs771056818 Chr.16: 3965904-3965904 on Build GRCh38
rs780415827 Chr.16: 3965913-3965913 on Build GRCh38
rs201541852 Chr.16: 3965906-3965906 on Build GRCh38
rs747632699 Chr.16: 3965847-3965847 on Build GRCh38
rs777551003 Chr.16: 3965991-3965991 on Build GRCh38
rs752248079 Chr.16: 3965917-3965917 on Build GRCh38
rs753965030 Chr.16: 3965892-3965892 on Build GRCh38
rs755792053 Chr.16: 3965922-3965922 on Build GRCh38
rs772806621 Chr.16: 3965852-3965852 on Build GRCh38
rs755711015 Chr.16: 3965988-3965988 on Build GRCh38
rs760974192 Chr.16: 3965885-3965885 on Build GRCh38
rs750878205 Chr.16: 3965971-3965971 on Build GRCh38
rs192788160 Chr.16: 3965941-3965941 on Build GRCh38
rs754619679 Chr.16: 3965843-3965843 on Build GRCh38
rs756879944 Chr.16: 3965994-3965994 on Build GRCh38
rs752922514 Chr.16: 3965922-3965923 on Build GRCh38
rs557051514 Chr.16: 3965850-3965850 on Build GRCh38
rs762822539 Chr.16: 3965849-3965852 on Build GRCh38
rs149834476 Chr.16: 3965869-3965869 on Build GRCh38
rs199644950 Chr.16: 3965833-3965833 on Build GRCh38
rs146633008 Chr.16: 3965940-3965940 on Build GRCh38
rs371323934 Chr.16: 3965824-3965824 on Build GRCh38
rs748891359 Chr.16: 3965932-3965932 on Build GRCh38
rs776569230 Chr.16: 3965957-3965957 on Build GRCh38
rs780861346 Chr.16: 3965844-3965844 on Build GRCh38
rs934867185 Chr.16: 3965925-3965925 on Build GRCh38
rs144464713 Chr.16: 3965905-3965905 on Build GRCh38
rs938200300 Chr.16: 3965910-3965910 on Build GRCh38
rs139504822 Chr.16: 3965811-3965811 on Build GRCh38
rs751963291 Chr.16: 3965982-3965982 on Build GRCh38
rs139771044 Chr.16: 3965883-3965883 on Build GRCh38
rs764041960 Chr.16: 3965891-3965891 on Build GRCh38
rs997578740 Chr.16: 3965849-3965849 on Build GRCh38
rs913886418 Chr.16: 3965900-3965900 on Build GRCh38
rs746289873 Chr.16: 3965835-3965835 on Build GRCh38
rs768979927 Chr.16: 3965953-3965953 on Build GRCh38
rs368305972 Chr.16: 3965938-3965938 on Build GRCh38
rs943523654 Chr.16: 3965977-3965977 on Build GRCh38
rs374081146 Chr.16: 3965834-3965834 on Build GRCh38
rs749110041 Chr.16: 3965858-3965858 on Build GRCh38
rs757814677 Chr.16: 3965818-3965818 on Build GRCh38
rs770771235 Chr.16: 3965936-3965936 on Build GRCh38
rs148729420 Chr.16: 3965947-3965947 on Build GRCh38
rs763480855 Chr.16: 3965978-3965978 on Build GRCh38
rs370863551 Chr.16: 3965979-3965979 on Build GRCh38
rs545884748 Chr.16: 3965895-3965895 on Build GRCh38
rs530150534 Chr.16: 3965960-3965960 on Build GRCh38

View Full Product Details