Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88733551-88733792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100289580
Entrez Gene ID
100289580
Gene Name
uncharacterized LOC100289580
Gene Aliases
-
UniGene
Hs.437712
Gene Chromosome Location
Chr.16: 88731180-88741425 on Build GRCh38
Transcripts
NR_103774.1 NM_001142864.3

Gene Symbol
PIEZO1
Entrez Gene ID
9780
Gene Name
piezo type mechanosensitive ion channel component 1
Gene Aliases
DHS, FAM38A, LMPH3, Mib
UniGene
Hs.377001
Gene Chromosome Location
Chr.16: 88715338-88785220 on Build GRCh38
Transcripts
NR_103774.1 NM_001142864.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5566836 c.2359C>T p.R787W Chr.16: 88733716-88733716 on Build GRCh38
4632136 c.2392G>A p.V798I Chr.16: 88733683-88733683 on Build GRCh38
5833206 c.2486_2487+1delAGG p.? Chr.16: 88733587-88733589 on Build GRCh38
5523467 c.2430G>A p.L810L Chr.16: 88733645-88733645 on Build GRCh38
7292714 c.2348T>C p.L783P Chr.16: 88733727-88733727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021969177 Chr.16: 88733674-88733674 on Build GRCh38
rs953601737 Chr.16: 88733738-88733738 on Build GRCh38
rs948699964 Chr.16: 88733622-88733622 on Build GRCh38
rs375353698 Chr.16: 88733672-88733672 on Build GRCh38
rs762333855 Chr.16: 88733751-88733751 on Build GRCh38
rs145574894 Chr.16: 88733569-88733569 on Build GRCh38
rs1015048100 Chr.16: 88733618-88733618 on Build GRCh38
rs781526622 Chr.16: 88733656-88733656 on Build GRCh38
rs931204966 Chr.16: 88733683-88733683 on Build GRCh38
rs990442192 Chr.16: 88733660-88733660 on Build GRCh38
rs1048314003 Chr.16: 88733626-88733626 on Build GRCh38
rs771820005 Chr.16: 88733581-88733581 on Build GRCh38
rs977657640 Chr.16: 88733700-88733700 on Build GRCh38
rs550646219 Chr.16: 88733772-88733772 on Build GRCh38
rs748012422 Chr.16: 88733687-88733687 on Build GRCh38
rs763268102 Chr.16: 88733710-88733710 on Build GRCh38
rs773008263 Chr.16: 88733707-88733707 on Build GRCh38
rs968995369 Chr.16: 88733688-88733688 on Build GRCh38
rs915946309 Chr.16: 88733717-88733717 on Build GRCh38
rs995448361 Chr.16: 88733597-88733597 on Build GRCh38
rs371733171 Chr.16: 88733771-88733771 on Build GRCh38
rs777786499 Chr.16: 88733676-88733677 on Build GRCh38
rs778818035 Chr.16: 88733684-88733684 on Build GRCh38
rs576808158 Chr.16: 88733654-88733654 on Build GRCh38
rs9940889 Chr.16: 88733761-88733761 on Build GRCh38
rs914794769 Chr.16: 88733584-88733584 on Build GRCh38
rs915932724 Chr.16: 88733620-88733620 on Build GRCh38
rs997631400 Chr.16: 88733730-88733730 on Build GRCh38
rs200970763 Chr.16: 88733731-88733731 on Build GRCh38
rs771739616 Chr.16: 88733695-88733695 on Build GRCh38
rs1004924007 Chr.16: 88733690-88733690 on Build GRCh38
rs760278106 Chr.16: 88733610-88733610 on Build GRCh38
rs774977877 Chr.16: 88733743-88733743 on Build GRCh38
rs960462384 Chr.16: 88733708-88733708 on Build GRCh38
rs763571494 Chr.16: 88733636-88733636 on Build GRCh38
rs563239344 Chr.16: 88733668-88733668 on Build GRCh38
rs9928479 Chr.16: 88733714-88733714 on Build GRCh38
rs367986919 Chr.16: 88733720-88733720 on Build GRCh38
rs542916104 Chr.16: 88733716-88733716 on Build GRCh38
rs986413987 Chr.16: 88733570-88733570 on Build GRCh38
rs563693664 Chr.16: 88733612-88733612 on Build GRCh38
rs370271938 Chr.16: 88733592-88733592 on Build GRCh38
rs901901180 Chr.16: 88733729-88733729 on Build GRCh38
rs563020910 Chr.16: 88733606-88733606 on Build GRCh38
rs924766274 Chr.16: 88733608-88733608 on Build GRCh38
rs888324060 Chr.16: 88733631-88733631 on Build GRCh38
rs1036856896 Chr.16: 88733643-88733643 on Build GRCh38
rs575029658 Chr.16: 88733653-88733653 on Build GRCh38
rs750634854 Chr.16: 88733665-88733665 on Build GRCh38
rs966196201 Chr.16: 88733619-88733619 on Build GRCh38
rs753985118 Chr.16: 88733770-88733770 on Build GRCh38
rs11649478 Chr.16: 88733768-88733768 on Build GRCh38
rs202103485 Chr.16: 88733652-88733652 on Build GRCh38
rs759116313 Chr.16: 88733728-88733728 on Build GRCh38
rs573397793 Chr.16: 88733624-88733624 on Build GRCh38
rs968879526 Chr.16: 88733596-88733596 on Build GRCh38
rs939967830 Chr.16: 88733767-88733767 on Build GRCh38
rs200029740 Chr.16: 88733666-88733666 on Build GRCh38
rs750230820 Chr.16: 88733680-88733680 on Build GRCh38
rs753334715 Chr.16: 88733635-88733635 on Build GRCh38
rs749307718 Chr.16: 88733671-88733671 on Build GRCh38
rs1031868414 Chr.16: 88733704-88733704 on Build GRCh38
rs1012235700 Chr.16: 88733746-88733747 on Build GRCh38
rs532138806 Chr.16: 88733769-88733769 on Build GRCh38
rs138064042 Chr.16: 88733609-88733609 on Build GRCh38
rs867405644 Chr.16: 88733676-88733676 on Build GRCh38

View Full Product Details