Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143992014-143992287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRINA
Entrez Gene ID
2907
Gene Name
glutamate ionotropic receptor NMDA type subunit associated protein 1
Gene Aliases
HNRGW, LFG1, NMDARA1, TMBIM3
UniGene
Hs.594634
Gene Chromosome Location
Chr.8: 143990058-143993415 on Build GRCh38
Transcripts
NM_000837.1 NM_001009184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2870523 c.636C>T p.I212I Chr.8: 143992021-143992021 on Build GRCh38
6842922 c.642C>A p.L214L Chr.8: 143992027-143992027 on Build GRCh38
5503433 c.693+8C>T p.? Chr.8: 143992086-143992086 on Build GRCh38
3898100 c.637G>A p.V213I Chr.8: 143992022-143992022 on Build GRCh38
1313846 c.707C>T p.A236V Chr.8: 143992258-143992258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782559819 Chr.8: 143992174-143992174 on Build GRCh38
rs782215676 Chr.8: 143992132-143992132 on Build GRCh38
rs782309962 Chr.8: 143992129-143992129 on Build GRCh38
rs782392953 Chr.8: 143992135-143992135 on Build GRCh38
rs371561711 Chr.8: 143992046-143992046 on Build GRCh38
rs782500571 Chr.8: 143992160-143992160 on Build GRCh38
rs781896785 Chr.8: 143992262-143992262 on Build GRCh38
rs116467837 Chr.8: 143992119-143992119 on Build GRCh38
rs782466380 Chr.8: 143992084-143992084 on Build GRCh38
rs372445018 Chr.8: 143992209-143992209 on Build GRCh38
rs373575365 Chr.8: 143992238-143992238 on Build GRCh38
rs1048241753 Chr.8: 143992144-143992144 on Build GRCh38
rs781906385 Chr.8: 143992050-143992050 on Build GRCh38
rs538211784 Chr.8: 143992265-143992265 on Build GRCh38
rs527950255 Chr.8: 143992191-143992191 on Build GRCh38
rs375429651 Chr.8: 143992214-143992214 on Build GRCh38
rs781947486 Chr.8: 143992149-143992149 on Build GRCh38
rs567074890 Chr.8: 143992188-143992188 on Build GRCh38
rs781956982 Chr.8: 143992252-143992253 on Build GRCh38
rs150522299 Chr.8: 143992247-143992247 on Build GRCh38
rs988390635 Chr.8: 143992042-143992042 on Build GRCh38
rs535765704 Chr.8: 143992175-143992175 on Build GRCh38
rs201648034 Chr.8: 143992113-143992113 on Build GRCh38
rs782539977 Chr.8: 143992114-143992114 on Build GRCh38
rs782553092 Chr.8: 143992066-143992066 on Build GRCh38
rs932374807 Chr.8: 143992139-143992139 on Build GRCh38
rs567639034 Chr.8: 143992216-143992216 on Build GRCh38
rs782687611 Chr.8: 143992257-143992257 on Build GRCh38
rs782471805 Chr.8: 143992181-143992181 on Build GRCh38
rs782023192 Chr.8: 143992197-143992197 on Build GRCh38
rs781883653 Chr.8: 143992169-143992169 on Build GRCh38
rs756808596 Chr.8: 143992131-143992131 on Build GRCh38
rs782451067 Chr.8: 143992246-143992246 on Build GRCh38
rs1002271584 Chr.8: 143992192-143992192 on Build GRCh38
rs781986924 Chr.8: 143992142-143992142 on Build GRCh38
rs201299903 Chr.8: 143992231-143992231 on Build GRCh38
rs532274486 Chr.8: 143992165-143992165 on Build GRCh38
rs782198762 Chr.8: 143992198-143992198 on Build GRCh38
rs782146869 Chr.8: 143992226-143992226 on Build GRCh38
rs781789043 Chr.8: 143992242-143992242 on Build GRCh38
rs958300501 Chr.8: 143992040-143992040 on Build GRCh38
rs958185393 Chr.8: 143992217-143992217 on Build GRCh38
rs782792940 Chr.8: 143992146-143992146 on Build GRCh38
rs782390497 Chr.8: 143992044-143992045 on Build GRCh38
rs782464392 Chr.8: 143992170-143992170 on Build GRCh38
rs782167163 Chr.8: 143992143-143992143 on Build GRCh38
rs781871359 Chr.8: 143992250-143992250 on Build GRCh38
rs781865845 Chr.8: 143992157-143992157 on Build GRCh38
rs781831117 Chr.8: 143992075-143992075 on Build GRCh38
rs1024948810 Chr.8: 143992179-143992179 on Build GRCh38
rs782251460 Chr.8: 143992189-143992189 on Build GRCh38
rs782296607 Chr.8: 143992256-143992256 on Build GRCh38
rs141499273 Chr.8: 143992215-143992215 on Build GRCh38
rs781944426 Chr.8: 143992210-143992210 on Build GRCh38
rs782592422 Chr.8: 143992072-143992072 on Build GRCh38
rs531227282 Chr.8: 143992091-143992091 on Build GRCh38

View Full Product Details