Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10559723-10559990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYVE1
Entrez Gene ID
10894
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Gene Aliases
CRSBP-1, HAR, LYVE-1, XLKD1
UniGene
Hs.655332
Gene Chromosome Location
Chr.11: 10557866-10568818 on Build GRCh38
Transcripts
NM_006691.3 NR_034093.2 NR_034094.2 NR_046374.1 NR_046375.1

Gene Symbol
MRVI1-AS1
Entrez Gene ID
100129827
Gene Name
MRVI1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.246769
Gene Chromosome Location
Chr.11: 10541236-10599932 on Build GRCh38
Transcripts
NM_006691.3 NR_034093.2 NR_034094.2 NR_046374.1 NR_046375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4507146 c.738C>T p.L246L Chr.11: 10559860-10559860 on Build GRCh38
4557117 c.715G>A p.A239T Chr.11: 10559883-10559883 on Build GRCh38
5792314 c.758G>C p.G253A Chr.11: 10559840-10559840 on Build GRCh38
4135664 c.781A>G p.R261G Chr.11: 10559817-10559817 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771243144 Chr.11: 10559885-10559885 on Build GRCh38
rs770674356 Chr.11: 10559784-10559784 on Build GRCh38
rs118170201 Chr.11: 10559890-10559890 on Build GRCh38
rs538173495 Chr.11: 10559787-10559787 on Build GRCh38
rs370002783 Chr.11: 10559779-10559779 on Build GRCh38
rs761919680 Chr.11: 10559950-10559950 on Build GRCh38
rs116575936 Chr.11: 10559810-10559810 on Build GRCh38
rs930123438 Chr.11: 10559801-10559801 on Build GRCh38
rs774758262 Chr.11: 10559887-10559887 on Build GRCh38
rs1006591910 Chr.11: 10559825-10559825 on Build GRCh38
rs371884025 Chr.11: 10559847-10559847 on Build GRCh38
rs533840110 Chr.11: 10559864-10559864 on Build GRCh38
rs765270056 Chr.11: 10559823-10559823 on Build GRCh38
rs749197330 Chr.11: 10559947-10559947 on Build GRCh38
rs12278495 Chr.11: 10559923-10559923 on Build GRCh38
rs549228485 Chr.11: 10559758-10559758 on Build GRCh38
rs961039092 Chr.11: 10559944-10559944 on Build GRCh38
rs764527381 Chr.11: 10559840-10559840 on Build GRCh38
rs748227692 Chr.11: 10559860-10559860 on Build GRCh38
rs768456049 Chr.11: 10559892-10559892 on Build GRCh38
rs140605904 Chr.11: 10559852-10559852 on Build GRCh38
rs779995039 Chr.11: 10559855-10559857 on Build GRCh38
rs771309216 Chr.11: 10559782-10559782 on Build GRCh38
rs779868511 Chr.11: 10559828-10559828 on Build GRCh38
rs569228657 Chr.11: 10559772-10559772 on Build GRCh38
rs151319034 Chr.11: 10559835-10559835 on Build GRCh38
rs761422124 Chr.11: 10559915-10559915 on Build GRCh38
rs368490518 Chr.11: 10559807-10559807 on Build GRCh38
rs750367525 Chr.11: 10559926-10559926 on Build GRCh38
rs770320641 Chr.11: 10559770-10559770 on Build GRCh38
rs758194425 Chr.11: 10559826-10559826 on Build GRCh38
rs755217678 Chr.11: 10559843-10559843 on Build GRCh38
rs367798406 Chr.11: 10559813-10559813 on Build GRCh38
rs960163171 Chr.11: 10559785-10559785 on Build GRCh38
rs117484197 Chr.11: 10559884-10559884 on Build GRCh38
rs778315152 Chr.11: 10559866-10559866 on Build GRCh38
rs1011426411 Chr.11: 10559867-10559867 on Build GRCh38
rs776357513 Chr.11: 10559911-10559911 on Build GRCh38
rs745985592 Chr.11: 10559817-10559817 on Build GRCh38
rs750361195 Chr.11: 10559824-10559824 on Build GRCh38
rs183625653 Chr.11: 10559750-10559750 on Build GRCh38

View Full Product Details