Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200893209-200893462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIF3L1
Entrez Gene ID
60491
Gene Name
NGG1 interacting factor 3 like 1
Gene Aliases
ALS2CR1, CALS-7, MDS015
UniGene
Hs.145284
Gene Chromosome Location
Chr.2: 200889327-200903932 on Build GRCh38
Transcripts
NM_001136039.2 NM_001142355.1 NM_001142356.1 NM_021824.3 XM_005246742.3 XM_011511582.1 XM_011511583.1 XM_011511584.1 XM_011511585.1 XM_017004630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1014776 c.366C>A p.T122T Chr.2: 200893256-200893256 on Build GRCh38
1014777 c.427G>T p.E143* Chr.2: 200893317-200893317 on Build GRCh38
6726123 c.490G>A p.G164S Chr.2: 200893380-200893380 on Build GRCh38
6561095 c.413G>A p.G138E Chr.2: 200893303-200893303 on Build GRCh38
4549436 c.472G>A p.A158T Chr.2: 200893281-200893281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775943124 Chr.2: 200893300-200893300 on Build GRCh38
rs368725799 Chr.2: 200893304-200893304 on Build GRCh38
rs959612952 Chr.2: 200893327-200893327 on Build GRCh38
rs778826172 Chr.2: 200893326-200893326 on Build GRCh38
rs752697169 Chr.2: 200893334-200893334 on Build GRCh38
rs760282968 Chr.2: 200893311-200893311 on Build GRCh38
rs374371539 Chr.2: 200893312-200893312 on Build GRCh38
rs747253244 Chr.2: 200893292-200893292 on Build GRCh38
rs978481619 Chr.2: 200893387-200893387 on Build GRCh38
rs967004456 Chr.2: 200893324-200893324 on Build GRCh38
rs368023292 Chr.2: 200893341-200893341 on Build GRCh38
rs745961449 Chr.2: 200893364-200893364 on Build GRCh38
rs777478902 Chr.2: 200893344-200893344 on Build GRCh38
rs751529842 Chr.2: 200893426-200893427 on Build GRCh38
rs781286405 Chr.2: 200893361-200893361 on Build GRCh38
rs759041195 Chr.2: 200893413-200893413 on Build GRCh38
rs770361317 Chr.2: 200893372-200893372 on Build GRCh38
rs763947372 Chr.2: 200893331-200893336 on Build GRCh38
rs749856115 Chr.2: 200893380-200893380 on Build GRCh38
rs760082371 Chr.2: 200893297-200893297 on Build GRCh38
rs375445740 Chr.2: 200893294-200893294 on Build GRCh38
rs771091763 Chr.2: 200893360-200893360 on Build GRCh38
rs776613414 Chr.2: 200893355-200893356 on Build GRCh38
rs776139756 Chr.2: 200893379-200893379 on Build GRCh38
rs147152104 Chr.2: 200893276-200893276 on Build GRCh38
rs201916129 Chr.2: 200893289-200893289 on Build GRCh38
rs776051945 Chr.2: 200893404-200893404 on Build GRCh38
rs760323226 Chr.2: 200893393-200893393 on Build GRCh38
rs753773806 Chr.2: 200893265-200893265 on Build GRCh38
rs141375721 Chr.2: 200893433-200893433 on Build GRCh38
rs371754717 Chr.2: 200893352-200893352 on Build GRCh38
rs77925669 Chr.2: 200893233-200893233 on Build GRCh38
rs774607467 Chr.2: 200893391-200893391 on Build GRCh38
rs764784737 Chr.2: 200893414-200893414 on Build GRCh38
rs371885073 Chr.2: 200893310-200893310 on Build GRCh38
rs367815278 Chr.2: 200893281-200893281 on Build GRCh38
rs758308750 Chr.2: 200893424-200893424 on Build GRCh38
rs200712116 Chr.2: 200893279-200893279 on Build GRCh38
rs776616197 Chr.2: 200893296-200893296 on Build GRCh38
rs761789873 Chr.2: 200893402-200893403 on Build GRCh38
rs752609063 Chr.2: 200893418-200893418 on Build GRCh38
rs770449083 Chr.2: 200893299-200893299 on Build GRCh38
rs560036003 Chr.2: 200893232-200893232 on Build GRCh38
rs760466709 Chr.2: 200893309-200893323 on Build GRCh38
rs754850747 Chr.2: 200893325-200893325 on Build GRCh38
rs769030817 Chr.2: 200893385-200893385 on Build GRCh38
rs754937497 Chr.2: 200893278-200893278 on Build GRCh38
rs765253260 Chr.2: 200893427-200893427 on Build GRCh38
rs375292044 Chr.2: 200893398-200893398 on Build GRCh38

View Full Product Details