Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41784868-41785141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRICKLE4
Entrez Gene ID
29964
Gene Name
prickle planar cell polarity protein 4
Gene Aliases
C6orf49, OBTP, OEBT, TOMM6
UniGene
Hs.715653
Gene Chromosome Location
Chr.6: 41780762-41787372 on Build GRCh38
Transcripts
NM_013397.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6755541 c.304C>T p.R102W Chr.6: 41784998-41784998 on Build GRCh38
484069 c.258C>T p.A86A Chr.6: 41784952-41784952 on Build GRCh38
6586903 c.288G>C p.Q96H Chr.6: 41784982-41784982 on Build GRCh38
6276728 c.343G>T p.V115L Chr.6: 41785037-41785037 on Build GRCh38
5792625 c.370T>C p.C124R Chr.6: 41785064-41785064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750454440 Chr.6: 41785076-41785076 on Build GRCh38
rs146832833 Chr.6: 41785031-41785031 on Build GRCh38
rs759909842 Chr.6: 41784950-41784951 on Build GRCh38
rs367669634 Chr.6: 41785025-41785025 on Build GRCh38
rs143845854 Chr.6: 41784999-41784999 on Build GRCh38
rs142658210 Chr.6: 41785032-41785032 on Build GRCh38
rs758480727 Chr.6: 41784921-41784921 on Build GRCh38
rs768142560 Chr.6: 41784973-41784973 on Build GRCh38
rs766830154 Chr.6: 41784907-41784907 on Build GRCh38
rs200826892 Chr.6: 41784969-41784969 on Build GRCh38
rs762985617 Chr.6: 41785119-41785119 on Build GRCh38
rs750751403 Chr.6: 41784953-41784953 on Build GRCh38
rs550794832 Chr.6: 41784940-41784940 on Build GRCh38
rs775372421 Chr.6: 41785020-41785020 on Build GRCh38
rs759660546 Chr.6: 41784913-41784913 on Build GRCh38
rs757171719 Chr.6: 41785034-41785034 on Build GRCh38
rs991801923 Chr.6: 41785097-41785097 on Build GRCh38
rs755743930 Chr.6: 41784958-41784958 on Build GRCh38
rs767099016 Chr.6: 41785023-41785027 on Build GRCh38
rs761191142 Chr.6: 41784905-41784905 on Build GRCh38
rs757294466 Chr.6: 41784938-41784938 on Build GRCh38
rs754310817 Chr.6: 41784911-41784911 on Build GRCh38
rs142771052 Chr.6: 41784939-41784939 on Build GRCh38
rs140175299 Chr.6: 41784992-41784992 on Build GRCh38
rs373105771 Chr.6: 41785105-41785105 on Build GRCh38
rs984424627 Chr.6: 41785004-41785004 on Build GRCh38
rs753025740 Chr.6: 41784916-41784916 on Build GRCh38
rs771348674 Chr.6: 41785093-41785093 on Build GRCh38
rs530736342 Chr.6: 41784893-41784893 on Build GRCh38
rs200288617 Chr.6: 41785078-41785078 on Build GRCh38
rs751283537 Chr.6: 41785020-41785021 on Build GRCh38
rs894856220 Chr.6: 41784923-41784923 on Build GRCh38
rs947739208 Chr.6: 41785109-41785109 on Build GRCh38
rs775672681 Chr.6: 41785115-41785115 on Build GRCh38
rs763262117 Chr.6: 41785027-41785027 on Build GRCh38
rs765213952 Chr.6: 41784915-41784915 on Build GRCh38
rs147741170 Chr.6: 41784968-41784968 on Build GRCh38
rs144794873 Chr.6: 41785087-41785087 on Build GRCh38
rs751352662 Chr.6: 41784932-41784932 on Build GRCh38
rs566145059 Chr.6: 41784998-41784998 on Build GRCh38
rs748704693 Chr.6: 41785089-41785089 on Build GRCh38
rs763961187 Chr.6: 41784924-41784924 on Build GRCh38
rs537237904 Chr.6: 41785040-41785040 on Build GRCh38
rs749920903 Chr.6: 41785087-41785088 on Build GRCh38
rs796307643 Chr.6: 41785020-41785021 on Build GRCh38
rs765361297 Chr.6: 41785012-41785012 on Build GRCh38
rs918415439 Chr.6: 41784975-41784975 on Build GRCh38
rs146162576 Chr.6: 41784974-41784974 on Build GRCh38
rs187184454 Chr.6: 41785092-41785092 on Build GRCh38
rs764215823 Chr.6: 41785121-41785121 on Build GRCh38
rs763727747 Chr.6: 41784958-41784960 on Build GRCh38
rs774787046 Chr.6: 41785015-41785015 on Build GRCh38
rs376424716 Chr.6: 41784980-41784980 on Build GRCh38
rs746384231 Chr.6: 41785095-41785095 on Build GRCh38
rs749275877 Chr.6: 41784996-41784996 on Build GRCh38

View Full Product Details