Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81002168-81002409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MESDC1
Entrez Gene ID
59274
Gene Name
mesoderm development candidate 1
Gene Aliases
-
UniGene
Hs.513071
Gene Chromosome Location
Chr.15: 81000954-81004004 on Build GRCh38
Transcripts
NM_022566.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7311009 c.48G>A p.P16P Chr.15: 81002319-81002319 on Build GRCh38
5714598 c.113A>C p.D38A Chr.15: 81002384-81002384 on Build GRCh38
4598144 c.101T>G p.V34G Chr.15: 81002372-81002372 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs918769138 Chr.15: 81002387-81002387 on Build GRCh38
rs999708388 Chr.15: 81002235-81002235 on Build GRCh38
rs1003557142 Chr.15: 81002201-81002201 on Build GRCh38
rs973342356 Chr.15: 81002356-81002356 on Build GRCh38
rs749627641 Chr.15: 81002383-81002383 on Build GRCh38
rs764640242 Chr.15: 81002304-81002304 on Build GRCh38
rs1013521120 Chr.15: 81002252-81002252 on Build GRCh38
rs746481339 Chr.15: 81002358-81002358 on Build GRCh38
rs940552952 Chr.15: 81002349-81002349 on Build GRCh38
rs551867821 Chr.15: 81002258-81002259 on Build GRCh38
rs906503274 Chr.15: 81002195-81002195 on Build GRCh38
rs201308091 Chr.15: 81002345-81002345 on Build GRCh38
rs530216587 Chr.15: 81002240-81002240 on Build GRCh38
rs542884250 Chr.15: 81002329-81002329 on Build GRCh38
rs1023280386 Chr.15: 81002254-81002254 on Build GRCh38
rs895394220 Chr.15: 81002388-81002388 on Build GRCh38
rs578200826 Chr.15: 81002315-81002315 on Build GRCh38
rs761884411 Chr.15: 81002327-81002327 on Build GRCh38
rs765079286 Chr.15: 81002343-81002343 on Build GRCh38
rs895400918 Chr.15: 81002204-81002204 on Build GRCh38
rs769397442 Chr.15: 81002371-81002371 on Build GRCh38
rs775909350 Chr.15: 81002372-81002372 on Build GRCh38
rs995469332 Chr.15: 81002194-81002211 on Build GRCh38
rs758357062 Chr.15: 81002348-81002348 on Build GRCh38
rs962893605 Chr.15: 81002352-81002352 on Build GRCh38
rs750636170 Chr.15: 81002333-81002333 on Build GRCh38
rs903677399 Chr.15: 81002189-81002189 on Build GRCh38
rs763508878 Chr.15: 81002298-81002298 on Build GRCh38
rs561230529 Chr.15: 81002210-81002210 on Build GRCh38
rs756845369 Chr.15: 81002361-81002361 on Build GRCh38
rs777861910 Chr.15: 81002351-81002351 on Build GRCh38
rs999803833 Chr.15: 81002198-81002198 on Build GRCh38
rs769083809 Chr.15: 81002391-81002391 on Build GRCh38
rs189162231 Chr.15: 81002335-81002335 on Build GRCh38
rs966069662 Chr.15: 81002289-81002289 on Build GRCh38
rs1022506715 Chr.15: 81002211-81002211 on Build GRCh38
rs745500291 Chr.15: 81002370-81002370 on Build GRCh38
rs748514254 Chr.15: 81002332-81002343 on Build GRCh38
rs966475272 Chr.15: 81002234-81002234 on Build GRCh38
rs1027494258 Chr.15: 81002203-81002214 on Build GRCh38
rs867429047 Chr.15: 81002375-81002375 on Build GRCh38

View Full Product Details