Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85665563-85665836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFTPB
Entrez Gene ID
6439
Gene Name
surfactant protein B
Gene Aliases
PSP-B, SFTB3, SFTP3, SMDP1, SP-B
UniGene
Hs.512690
Gene Chromosome Location
Chr.2: 85657317-85668741 on Build GRCh38
Transcripts
NM_000542.3 NM_198843.2 XM_005264487.2 XM_005264488.3 XM_005264490.4 XM_017004702.1 XM_017004703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7154804 c.529G>A p.V177I Chr.2: 85665659-85665659 on Build GRCh38
1690758 c.496G>A p.D166N Chr.2: 85665692-85665692 on Build GRCh38
39342 c.532C>T p.L178F Chr.2: 85665656-85665656 on Build GRCh38
3963181 c.474C>A p.D158E Chr.2: 85665714-85665714 on Build GRCh38
263617 c.544C>T p.P182S Chr.2: 85665644-85665644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370715446 Chr.2: 85665695-85665695 on Build GRCh38
rs768712184 Chr.2: 85665713-85665713 on Build GRCh38
rs370750455 Chr.2: 85665694-85665694 on Build GRCh38
rs751412409 Chr.2: 85665776-85665776 on Build GRCh38
rs982427444 Chr.2: 85665675-85665675 on Build GRCh38
rs927636207 Chr.2: 85665597-85665597 on Build GRCh38
rs749997825 Chr.2: 85665638-85665638 on Build GRCh38
rs752321943 Chr.2: 85665768-85665768 on Build GRCh38
rs529554098 Chr.2: 85665792-85665792 on Build GRCh38
rs200635375 Chr.2: 85665590-85665590 on Build GRCh38
rs184494733 Chr.2: 85665641-85665641 on Build GRCh38
rs374131015 Chr.2: 85665786-85665786 on Build GRCh38
rs548370069 Chr.2: 85665668-85665668 on Build GRCh38
rs754507327 Chr.2: 85665663-85665663 on Build GRCh38
rs748780640 Chr.2: 85665608-85665608 on Build GRCh38
rs761127200 Chr.2: 85665811-85665811 on Build GRCh38
rs947459095 Chr.2: 85665765-85665765 on Build GRCh38
rs138274558 Chr.2: 85665644-85665644 on Build GRCh38
rs149904344 Chr.2: 85665678-85665678 on Build GRCh38
rs778119902 Chr.2: 85665638-85665638 on Build GRCh38
rs35373464 Chr.2: 85665785-85665785 on Build GRCh38
rs45601634 Chr.2: 85665612-85665612 on Build GRCh38
rs375203471 Chr.2: 85665709-85665709 on Build GRCh38
rs767972982 Chr.2: 85665593-85665593 on Build GRCh38
rs756571077 Chr.2: 85665628-85665628 on Build GRCh38
rs748156830 Chr.2: 85665807-85665807 on Build GRCh38
rs773147825 Chr.2: 85665624-85665624 on Build GRCh38
rs549877442 Chr.2: 85665766-85665766 on Build GRCh38
rs764711051 Chr.2: 85665590-85665591 on Build GRCh38
rs777381019 Chr.2: 85665775-85665775 on Build GRCh38
rs773765909 Chr.2: 85665729-85665729 on Build GRCh38
rs1030295362 Chr.2: 85665653-85665653 on Build GRCh38
rs3024801 Chr.2: 85665662-85665662 on Build GRCh38
rs773567727 Chr.2: 85665810-85665810 on Build GRCh38
rs767932734 Chr.2: 85665645-85665645 on Build GRCh38
rs886044577 Chr.2: 85665600-85665600 on Build GRCh38
rs772485307 Chr.2: 85665808-85665808 on Build GRCh38
rs45505791 Chr.2: 85665642-85665642 on Build GRCh38
rs762058470 Chr.2: 85665714-85665714 on Build GRCh38
rs531280852 Chr.2: 85665752-85665752 on Build GRCh38
rs771834627 Chr.2: 85665619-85665619 on Build GRCh38
rs778434631 Chr.2: 85665669-85665669 on Build GRCh38
rs35524245 Chr.2: 85665640-85665640 on Build GRCh38
rs770903531 Chr.2: 85665700-85665700 on Build GRCh38
rs781094026 Chr.2: 85665703-85665703 on Build GRCh38
rs937667239 Chr.2: 85665798-85665798 on Build GRCh38
rs1019814885 Chr.2: 85665629-85665629 on Build GRCh38
rs367831550 Chr.2: 85665751-85665751 on Build GRCh38
rs377545629 Chr.2: 85665616-85665616 on Build GRCh38
rs1026496084 Chr.2: 85665762-85665762 on Build GRCh38
rs755711266 Chr.2: 85665601-85665601 on Build GRCh38
rs779673668 Chr.2: 85665603-85665603 on Build GRCh38
rs750890243 Chr.2: 85665596-85665596 on Build GRCh38
rs377624872 Chr.2: 85665672-85665672 on Build GRCh38
rs779093128 Chr.2: 85665806-85665806 on Build GRCh38
rs935119498 Chr.2: 85665701-85665701 on Build GRCh38
rs141123597 Chr.2: 85665618-85665618 on Build GRCh38
rs778541429 Chr.2: 85665716-85665716 on Build GRCh38
rs34550459 Chr.2: 85665659-85665659 on Build GRCh38
rs142376116 Chr.2: 85665588-85665589 on Build GRCh38
rs147209702 Chr.2: 85665660-85665660 on Build GRCh38
rs543913426 Chr.2: 85665627-85665627 on Build GRCh38
rs751001412 Chr.2: 85665657-85665657 on Build GRCh38
rs768637269 Chr.2: 85665800-85665800 on Build GRCh38
rs758084893 Chr.2: 85665771-85665771 on Build GRCh38

View Full Product Details