Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 170717321-170717568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101926913
Entrez Gene ID
101926913
Gene Name
uncharacterized LOC101926913
Gene Aliases
-
UniGene
Hs.301715
Gene Chromosome Location
Chr.2: 170700368-170770766 on Build GRCh38
Transcripts
NR_110185.1 NM_001003845.2 XM_005246542.4 XM_011511159.2

Gene Symbol
SP5
Entrez Gene ID
389058
Gene Name
Sp5 transcription factor
Gene Aliases
-
UniGene
Hs.368802
Gene Chromosome Location
Chr.2: 170714725-170717988 on Build GRCh38
Transcripts
NR_110185.1 NM_001003845.2 XM_005246542.4 XM_011511159.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5486189 c.1156G>A p.A386T Chr.2: 170717363-170717363 on Build GRCh38
4086707 c.1184C>T p.A395V Chr.2: 170717391-170717391 on Build GRCh38
6893641 c.1169T>C p.V390A Chr.2: 170717376-170717376 on Build GRCh38
1227201 c.1186C>T p.R396W Chr.2: 170717393-170717393 on Build GRCh38
7391511 c.1156G>T p.A386S Chr.2: 170717363-170717363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781706380 Chr.2: 170717400-170717400 on Build GRCh38
rs1043003543 Chr.2: 170717499-170717499 on Build GRCh38
rs201465830 Chr.2: 170717373-170717373 on Build GRCh38
rs1012011796 Chr.2: 170717344-170717344 on Build GRCh38
rs572881879 Chr.2: 170717346-170717346 on Build GRCh38
rs577072543 Chr.2: 170717471-170717471 on Build GRCh38
rs75866412 Chr.2: 170717417-170717417 on Build GRCh38
rs535169655 Chr.2: 170717436-170717436 on Build GRCh38
rs748701511 Chr.2: 170717403-170717403 on Build GRCh38
rs770340130 Chr.2: 170717348-170717350 on Build GRCh38
rs766846978 Chr.2: 170717430-170717432 on Build GRCh38
rs954508294 Chr.2: 170717530-170717530 on Build GRCh38
rs538142152 Chr.2: 170717504-170717504 on Build GRCh38
rs1056363347 Chr.2: 170717546-170717546 on Build GRCh38
rs193297268 Chr.2: 170717449-170717449 on Build GRCh38
rs909293842 Chr.2: 170717526-170717526 on Build GRCh38
rs773798827 Chr.2: 170717421-170717421 on Build GRCh38
rs773654556 Chr.2: 170717412-170717412 on Build GRCh38
rs750571745 Chr.2: 170717364-170717364 on Build GRCh38
rs770749676 Chr.2: 170717426-170717426 on Build GRCh38
rs758670097 Chr.2: 170717370-170717370 on Build GRCh38
rs775224848 Chr.2: 170717454-170717454 on Build GRCh38
rs979999004 Chr.2: 170717382-170717382 on Build GRCh38
rs1011218414 Chr.2: 170717462-170717462 on Build GRCh38
rs73015354 Chr.2: 170717351-170717351 on Build GRCh38
rs776089505 Chr.2: 170717502-170717502 on Build GRCh38
rs558703157 Chr.2: 170717455-170717455 on Build GRCh38
rs773996561 Chr.2: 170717433-170717433 on Build GRCh38
rs755470815 Chr.2: 170717389-170717389 on Build GRCh38
rs929897641 Chr.2: 170717542-170717542 on Build GRCh38
rs36058157 Chr.2: 170717501-170717502 on Build GRCh38
rs747129605 Chr.2: 170717381-170717381 on Build GRCh38
rs769981602 Chr.2: 170717405-170717405 on Build GRCh38
rs201188860 Chr.2: 170717363-170717363 on Build GRCh38
rs1038592341 Chr.2: 170717528-170717528 on Build GRCh38
rs896965682 Chr.2: 170717535-170717535 on Build GRCh38
rs996060075 Chr.2: 170717503-170717503 on Build GRCh38
rs957159049 Chr.2: 170717420-170717420 on Build GRCh38
rs143436539 Chr.2: 170717527-170717527 on Build GRCh38
rs796067095 Chr.2: 170717450-170717450 on Build GRCh38
rs763469321 Chr.2: 170717421-170717422 on Build GRCh38
rs185558675 Chr.2: 170717519-170717519 on Build GRCh38

View Full Product Details