Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12668951-12669212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR83
Entrez Gene ID
84292
Gene Name
WD repeat domain 83
Gene Aliases
MORG1
UniGene
Hs.657204
Gene Chromosome Location
Chr.19: 12666804-12675832 on Build GRCh38
Transcripts
NM_001099737.2 NR_029375.1 NM_016145.3 XM_017026864.1

Gene Symbol
WDR83OS
Entrez Gene ID
51398
Gene Name
WD repeat domain 83 opposite strand
Gene Aliases
C19orf56, PTD008
UniGene
Hs.108969
Gene Chromosome Location
Chr.19: 12668067-12671023 on Build GRCh38
Transcripts
NM_001099737.2 NR_029375.1 NM_016145.3 XM_017026864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5505640 c.152T>G p.L51R Chr.19: 12669132-12669132 on Build GRCh38
4790213 c.124A>G p.M42V Chr.19: 12669160-12669160 on Build GRCh38
4937944 c.102G>A p.P34P Chr.19: 12669182-12669182 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780132258 Chr.19: 12669096-12669096 on Build GRCh38
rs564388688 Chr.19: 12669118-12669118 on Build GRCh38
rs181324830 Chr.19: 12669092-12669092 on Build GRCh38
rs1020705935 Chr.19: 12669177-12669177 on Build GRCh38
rs748025928 Chr.19: 12669142-12669142 on Build GRCh38
rs373186687 Chr.19: 12669078-12669078 on Build GRCh38
rs1042480204 Chr.19: 12668996-12668996 on Build GRCh38
rs575346683 Chr.19: 12669107-12669107 on Build GRCh38
rs371115950 Chr.19: 12669079-12669079 on Build GRCh38
rs202078147 Chr.19: 12669169-12669169 on Build GRCh38
rs764626242 Chr.19: 12669077-12669077 on Build GRCh38
rs369956598 Chr.19: 12668983-12668983 on Build GRCh38
rs769701436 Chr.19: 12669149-12669149 on Build GRCh38
rs779140308 Chr.19: 12669085-12669085 on Build GRCh38
rs545848666 Chr.19: 12669113-12669113 on Build GRCh38
rs762685440 Chr.19: 12669150-12669150 on Build GRCh38
rs752884467 Chr.19: 12669015-12669015 on Build GRCh38
rs374487757 Chr.19: 12669095-12669095 on Build GRCh38
rs759056832 Chr.19: 12669166-12669166 on Build GRCh38
rs1049342930 Chr.19: 12669024-12669024 on Build GRCh38
rs1009357198 Chr.19: 12669128-12669128 on Build GRCh38
rs143946475 Chr.19: 12669046-12669049 on Build GRCh38
rs535503979 Chr.19: 12668979-12668979 on Build GRCh38
rs762941858 Chr.19: 12669071-12669071 on Build GRCh38
rs762195782 Chr.19: 12669175-12669175 on Build GRCh38
rs754433938 Chr.19: 12669170-12669170 on Build GRCh38
rs774000365 Chr.19: 12669161-12669161 on Build GRCh38
rs770663847 Chr.19: 12669157-12669157 on Build GRCh38
rs998236839 Chr.19: 12669049-12669049 on Build GRCh38

View Full Product Details