Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 25004705-25004940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARX
Entrez Gene ID
170302
Gene Name
aristaless related homeobox
Gene Aliases
CT121, EIEE1, ISSX, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS
UniGene
Hs.300304
Gene Chromosome Location
Chr.X: 25003694-25015948 on Build GRCh38
Transcripts
NM_139058.2

SNP Detail

SNP ID Chromosome Location
rs938274319 Chr.X: 25004738-25004738 on Build GRCh38
rs752807804 Chr.X: 25004888-25004888 on Build GRCh38
rs900885863 Chr.X: 25004831-25004831 on Build GRCh38
rs759074085 Chr.X: 25004896-25004896 on Build GRCh38
rs775559473 Chr.X: 25004808-25004808 on Build GRCh38
rs766065019 Chr.X: 25004884-25004884 on Build GRCh38
rs1056460685 Chr.X: 25004835-25004835 on Build GRCh38
rs896527800 Chr.X: 25004836-25004836 on Build GRCh38
rs747042039 Chr.X: 25004813-25004813 on Build GRCh38
rs762710900 Chr.X: 25004879-25004879 on Build GRCh38
rs752649243 Chr.X: 25004909-25004909 on Build GRCh38
rs777179729 Chr.X: 25004838-25004838 on Build GRCh38
rs770318189 Chr.X: 25004862-25004862 on Build GRCh38
rs773666007 Chr.X: 25004871-25004871 on Build GRCh38
rs777566066 Chr.X: 25004916-25004916 on Build GRCh38
rs746120093 Chr.X: 25004798-25004798 on Build GRCh38
rs398124509 Chr.X: 25004844-25004844 on Build GRCh38
rs1057517705 Chr.X: 25004759-25004760 on Build GRCh38
rs387906715 Chr.X: 25004755-25004755 on Build GRCh38
rs769107210 Chr.X: 25004829-25004829 on Build GRCh38
rs767404024 Chr.X: 25004897-25004897 on Build GRCh38
rs797045292 Chr.X: 25004887-25004888 on Build GRCh38
rs587783191 Chr.X: 25004894-25004894 on Build GRCh38
rs755881928 Chr.X: 25004731-25004731 on Build GRCh38
rs1057518794 Chr.X: 25004777-25004780 on Build GRCh38
rs747952341 Chr.X: 25004730-25004730 on Build GRCh38
rs587783192 Chr.X: 25004815-25004815 on Build GRCh38
rs772231944 Chr.X: 25004802-25004802 on Build GRCh38
rs779145937 Chr.X: 25004796-25004796 on Build GRCh38
rs1021002740 Chr.X: 25004868-25004868 on Build GRCh38

View Full Product Details