Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64521805-64522052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102723809
Entrez Gene ID
102723809
Gene Name
uncharacterized LOC102723809
Gene Aliases
-
UniGene
Hs.667192
Gene Chromosome Location
Chr.14: 64514151-64540368 on Build GRCh38
Transcripts
NR_110550.1 NM_001123329.1 NM_014950.2 XM_005267410.1 XM_011536566.2 XM_011536568.2 XM_017021094.1 XM_017021095.1

Gene Symbol
ZBTB1
Entrez Gene ID
22890
Gene Name
zinc finger and BTB domain containing 1
Gene Aliases
ZNF909
UniGene
Hs.605143
Gene Chromosome Location
Chr.14: 64504574-64533690 on Build GRCh38
Transcripts
NR_110550.1 NM_001123329.1 NM_014950.2 XM_005267410.1 XM_011536566.2 XM_011536568.2 XM_017021094.1 XM_017021095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5634139 c.301A>G p.M101V Chr.14: 64521805-64521805 on Build GRCh38
3377503 c.373A>G p.K125E Chr.14: 64521877-64521877 on Build GRCh38
4051806 c.312A>G p.L104L Chr.14: 64521816-64521816 on Build GRCh38
5788266 c.431T>G p.M144R Chr.14: 64521935-64521935 on Build GRCh38
4437102 c.486G>A p.E162E Chr.14: 64521990-64521990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762114031 Chr.14: 64521866-64521866 on Build GRCh38
rs376571289 Chr.14: 64521983-64521983 on Build GRCh38
rs769530319 Chr.14: 64521948-64521948 on Build GRCh38
rs777471462 Chr.14: 64521955-64521955 on Build GRCh38
rs749008388 Chr.14: 64521966-64521966 on Build GRCh38
rs751692094 Chr.14: 64521887-64521887 on Build GRCh38
rs944898727 Chr.14: 64521995-64521995 on Build GRCh38
rs1025058811 Chr.14: 64521891-64521891 on Build GRCh38
rs895241853 Chr.14: 64521863-64521863 on Build GRCh38
rs775972428 Chr.14: 64521845-64521845 on Build GRCh38
rs910679956 Chr.14: 64521987-64521987 on Build GRCh38
rs978483191 Chr.14: 64521919-64521919 on Build GRCh38
rs1040562978 Chr.14: 64522010-64522010 on Build GRCh38
rs770486534 Chr.14: 64521841-64521841 on Build GRCh38
rs970774761 Chr.14: 64521912-64521912 on Build GRCh38
rs762215264 Chr.14: 64522016-64522016 on Build GRCh38
rs761105445 Chr.14: 64521846-64521846 on Build GRCh38
rs770574476 Chr.14: 64521967-64521967 on Build GRCh38
rs773794184 Chr.14: 64521973-64521973 on Build GRCh38
rs542550426 Chr.14: 64521864-64521864 on Build GRCh38
rs780344438 Chr.14: 64521885-64521885 on Build GRCh38
rs765574760 Chr.14: 64522023-64522023 on Build GRCh38
rs1026782070 Chr.14: 64521850-64521850 on Build GRCh38
rs777044118 Chr.14: 64522003-64522003 on Build GRCh38
rs753507746 Chr.14: 64521928-64521928 on Build GRCh38
rs758535582 Chr.14: 64521884-64521884 on Build GRCh38
rs371466604 Chr.14: 64521880-64521880 on Build GRCh38
rs746132771 Chr.14: 64521848-64521849 on Build GRCh38
rs747947004 Chr.14: 64521945-64521945 on Build GRCh38
rs1050553649 Chr.14: 64521934-64521934 on Build GRCh38
rs188036695 Chr.14: 64522002-64522002 on Build GRCh38
rs150032061 Chr.14: 64521838-64521838 on Build GRCh38
rs573689136 Chr.14: 64521857-64521857 on Build GRCh38
rs930705753 Chr.14: 64521855-64521855 on Build GRCh38
rs1012369488 Chr.14: 64521865-64521865 on Build GRCh38
rs541812701 Chr.14: 64521867-64521867 on Build GRCh38
rs781025755 Chr.14: 64521938-64521938 on Build GRCh38

View Full Product Details