Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38306173-38306420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A14
Entrez Gene ID
9389
Gene Name
solute carrier family 22 member 14
Gene Aliases
OCTL2, OCTL4, ORCTL4
UniGene
Hs.165559
Gene Chromosome Location
Chr.3: 38281662-38318575 on Build GRCh38
Transcripts
NM_001320033.1 NM_004803.4 XM_005265585.4 XM_006713416.3 XM_006713417.3 XM_006713418.3 XM_011534245.2 XM_011534247.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
446438 c.334A>G p.K112E Chr.3: 38306360-38306360 on Build GRCh38
6251148 c.188G>T p.G63V Chr.3: 38306214-38306214 on Build GRCh38
1485127 c.265T>A p.F89I Chr.3: 38306291-38306291 on Build GRCh38
3660514 c.266T>C p.F89S Chr.3: 38306292-38306292 on Build GRCh38
5399213 c.225C>T p.I75I Chr.3: 38306251-38306251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748915002 Chr.3: 38306199-38306199 on Build GRCh38
rs369896741 Chr.3: 38306230-38306230 on Build GRCh38
rs756029665 Chr.3: 38306293-38306293 on Build GRCh38
rs776114995 Chr.3: 38306210-38306210 on Build GRCh38
rs761948692 Chr.3: 38306253-38306253 on Build GRCh38
rs756007782 Chr.3: 38306394-38306394 on Build GRCh38
rs769143324 Chr.3: 38306357-38306357 on Build GRCh38
rs566174873 Chr.3: 38306392-38306392 on Build GRCh38
rs775094730 Chr.3: 38306346-38306346 on Build GRCh38
rs764392380 Chr.3: 38306225-38306225 on Build GRCh38
rs773393219 Chr.3: 38306387-38306387 on Build GRCh38
rs9847584 Chr.3: 38306349-38306349 on Build GRCh38
rs147752298 Chr.3: 38306294-38306294 on Build GRCh38
rs772761302 Chr.3: 38306206-38306206 on Build GRCh38
rs145815885 Chr.3: 38306345-38306345 on Build GRCh38
rs1001339019 Chr.3: 38306305-38306305 on Build GRCh38
rs750561863 Chr.3: 38306266-38306266 on Build GRCh38
rs763123196 Chr.3: 38306388-38306388 on Build GRCh38
rs930909465 Chr.3: 38306221-38306221 on Build GRCh38
rs745364463 Chr.3: 38306205-38306205 on Build GRCh38
rs905649969 Chr.3: 38306302-38306302 on Build GRCh38
rs1021466759 Chr.3: 38306256-38306266 on Build GRCh38
rs966688999 Chr.3: 38306369-38306369 on Build GRCh38
rs765429335 Chr.3: 38306384-38306384 on Build GRCh38
rs879593528 Chr.3: 38306393-38306393 on Build GRCh38
rs758451137 Chr.3: 38306271-38306271 on Build GRCh38
rs963045791 Chr.3: 38306259-38306259 on Build GRCh38
rs929912931 Chr.3: 38306288-38306288 on Build GRCh38
rs760278011 Chr.3: 38306262-38306263 on Build GRCh38
rs778692901 Chr.3: 38306327-38306327 on Build GRCh38
rs2073714 Chr.3: 38306360-38306360 on Build GRCh38
rs9847670 Chr.3: 38306203-38306203 on Build GRCh38
rs752661992 Chr.3: 38306277-38306277 on Build GRCh38
rs765494260 Chr.3: 38306265-38306265 on Build GRCh38
rs745415733 Chr.3: 38306343-38306343 on Build GRCh38
rs372904499 Chr.3: 38306319-38306319 on Build GRCh38
rs200420643 Chr.3: 38306200-38306200 on Build GRCh38
rs764086331 Chr.3: 38306395-38306395 on Build GRCh38
rs749044560 Chr.3: 38306295-38306295 on Build GRCh38
rs762225721 Chr.3: 38306365-38306365 on Build GRCh38
rs373698627 Chr.3: 38306358-38306358 on Build GRCh38
rs776999841 Chr.3: 38306251-38306251 on Build GRCh38
rs145348200 Chr.3: 38306220-38306220 on Build GRCh38
rs767484064 Chr.3: 38306272-38306272 on Build GRCh38

View Full Product Details