Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60757444-60757717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL7
Entrez Gene ID
79993
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
-
UniGene
Hs.274256
Gene Chromosome Location
Chr.5: 60751789-60844292 on Build GRCh38
Transcripts
NM_001104558.1 NM_001297617.1 NM_001297618.1 NM_024930.2 XM_005248606.4 XM_005248607.3 XM_006714695.3 XM_011543651.2 XM_017009885.1 XM_017009886.1 XM_017009887.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
482935 c.514T>C p.F172L Chr.5: 60757631-60757631 on Build GRCh38
6326464 c.536C>T p.A179V Chr.5: 60757609-60757609 on Build GRCh38
2153112 c.509G>A p.G170E Chr.5: 60757636-60757636 on Build GRCh38
325250 c.606G>T p.W202C Chr.5: 60757539-60757539 on Build GRCh38
6104468 c.580G>T p.G194W Chr.5: 60757565-60757565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538889611 Chr.5: 60757685-60757685 on Build GRCh38
rs761698111 Chr.5: 60757582-60757582 on Build GRCh38
rs748514510 Chr.5: 60757511-60757511 on Build GRCh38
rs372397313 Chr.5: 60757603-60757603 on Build GRCh38
rs766584906 Chr.5: 60757607-60757607 on Build GRCh38
rs769502101 Chr.5: 60757544-60757544 on Build GRCh38
rs755504708 Chr.5: 60757481-60757481 on Build GRCh38
rs748788441 Chr.5: 60757672-60757672 on Build GRCh38
rs200475308 Chr.5: 60757533-60757533 on Build GRCh38
rs769625263 Chr.5: 60757664-60757664 on Build GRCh38
rs887860206 Chr.5: 60757648-60757648 on Build GRCh38
rs1040608592 Chr.5: 60757554-60757554 on Build GRCh38
rs773675782 Chr.5: 60757528-60757528 on Build GRCh38
rs570014111 Chr.5: 60757617-60757617 on Build GRCh38
rs763982943 Chr.5: 60757573-60757573 on Build GRCh38
rs375469282 Chr.5: 60757625-60757625 on Build GRCh38
rs774313366 Chr.5: 60757577-60757577 on Build GRCh38
rs761039870 Chr.5: 60757582-60757582 on Build GRCh38
rs139703859 Chr.5: 60757568-60757568 on Build GRCh38
rs1022187545 Chr.5: 60757483-60757483 on Build GRCh38
rs567565911 Chr.5: 60757521-60757521 on Build GRCh38
rs529994057 Chr.5: 60757563-60757563 on Build GRCh38
rs779731231 Chr.5: 60757669-60757669 on Build GRCh38
rs531969561 Chr.5: 60757484-60757484 on Build GRCh38
rs758928464 Chr.5: 60757629-60757629 on Build GRCh38
rs779332363 Chr.5: 60757622-60757622 on Build GRCh38
rs764298261 Chr.5: 60757622-60757622 on Build GRCh38
rs753924328 Chr.5: 60757614-60757614 on Build GRCh38
rs747370460 Chr.5: 60757661-60757661 on Build GRCh38
rs767320792 Chr.5: 60757596-60757596 on Build GRCh38
rs768244001 Chr.5: 60757673-60757673 on Build GRCh38

View Full Product Details