Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119715541-119715774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAATS1
Entrez Gene ID
89876
Gene Name
MYCBP associated and testis expressed 1
Gene Aliases
AAT1, AAT1alpha, C3orf15, CaM-IP2, SPATA26
UniGene
Hs.341906
Gene Chromosome Location
Chr.3: 119703022-119767102 on Build GRCh38
Transcripts
NM_001320316.1 NM_001320317.1 NM_001320318.1 NM_033364.3 XM_011513277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3914637 c.545C>T p.S182F Chr.3: 119715606-119715606 on Build GRCh38
4849720 c.657C>T p.L219L Chr.3: 119715718-119715718 on Build GRCh38
7151560 c.621A>G p.A207A Chr.3: 119715682-119715682 on Build GRCh38
4941665 c.567G>A p.V189V Chr.3: 119715628-119715628 on Build GRCh38
5417603 c.615C>T p.Y205Y Chr.3: 119715676-119715676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150795242 Chr.3: 119715579-119715579 on Build GRCh38
rs6438544 Chr.3: 119715680-119715680 on Build GRCh38
rs753051761 Chr.3: 119715733-119715733 on Build GRCh38
rs764184030 Chr.3: 119715604-119715604 on Build GRCh38
rs754360079 Chr.3: 119715710-119715710 on Build GRCh38
rs777243856 Chr.3: 119715637-119715637 on Build GRCh38
rs565776294 Chr.3: 119715659-119715659 on Build GRCh38
rs568392472 Chr.3: 119715707-119715707 on Build GRCh38
rs745984396 Chr.3: 119715749-119715749 on Build GRCh38
rs779207550 Chr.3: 119715609-119715609 on Build GRCh38
rs147597066 Chr.3: 119715744-119715744 on Build GRCh38
rs550236089 Chr.3: 119715695-119715695 on Build GRCh38
rs750575398 Chr.3: 119715587-119715587 on Build GRCh38
rs1041983496 Chr.3: 119715658-119715658 on Build GRCh38
rs1055729842 Chr.3: 119715600-119715600 on Build GRCh38
rs751602858 Chr.3: 119715617-119715617 on Build GRCh38
rs758785479 Chr.3: 119715739-119715739 on Build GRCh38
rs773596379 Chr.3: 119715688-119715688 on Build GRCh38
rs951168502 Chr.3: 119715573-119715573 on Build GRCh38
rs751791444 Chr.3: 119715576-119715576 on Build GRCh38
rs765608012 Chr.3: 119715732-119715732 on Build GRCh38
rs373361814 Chr.3: 119715648-119715648 on Build GRCh38
rs199907538 Chr.3: 119715673-119715673 on Build GRCh38
rs769625913 Chr.3: 119715672-119715672 on Build GRCh38
rs560430820 Chr.3: 119715628-119715628 on Build GRCh38
rs977019196 Chr.3: 119715667-119715667 on Build GRCh38
rs776492447 Chr.3: 119715650-119715650 on Build GRCh38
rs139064408 Chr.3: 119715675-119715675 on Build GRCh38
rs914441437 Chr.3: 119715626-119715626 on Build GRCh38
rs936985209 Chr.3: 119715596-119715596 on Build GRCh38
rs369945433 Chr.3: 119715586-119715586 on Build GRCh38
rs201993047 Chr.3: 119715686-119715686 on Build GRCh38
rs755506894 Chr.3: 119715725-119715725 on Build GRCh38
rs145890924 Chr.3: 119715594-119715594 on Build GRCh38
rs1027064201 Chr.3: 119715580-119715580 on Build GRCh38
rs770982721 Chr.3: 119715691-119715691 on Build GRCh38
rs756281021 Chr.3: 119715589-119715589 on Build GRCh38
rs746475787 Chr.3: 119715647-119715647 on Build GRCh38

View Full Product Details