Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29136979-29137242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A1
Entrez Gene ID
23359
Gene Name
family with sequence similarity 189 member A1
Gene Aliases
TMEM228
UniGene
Hs.383564
Gene Chromosome Location
Chr.15: 29120252-29570723 on Build GRCh38
Transcripts
NM_015307.1 XM_011521407.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7367235 c.646T>C p.S216P Chr.15: 29137179-29137179 on Build GRCh38
5383123 c.744C>T p.P248P Chr.15: 29137081-29137081 on Build GRCh38
4639397 c.688G>A p.V230I Chr.15: 29137137-29137137 on Build GRCh38
3956522 c.753C>A p.P251P Chr.15: 29137072-29137072 on Build GRCh38
5383122 c.750C>T p.Y250Y Chr.15: 29137075-29137075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959217761 Chr.15: 29137181-29137181 on Build GRCh38
rs576368016 Chr.15: 29137138-29137138 on Build GRCh38
rs61741714 Chr.15: 29137144-29137144 on Build GRCh38
rs769245050 Chr.15: 29137057-29137057 on Build GRCh38
rs761625316 Chr.15: 29137160-29137160 on Build GRCh38
rs981891721 Chr.15: 29137155-29137155 on Build GRCh38
rs754824223 Chr.15: 29137075-29137075 on Build GRCh38
rs935031635 Chr.15: 29137085-29137085 on Build GRCh38
rs947949852 Chr.15: 29137026-29137026 on Build GRCh38
rs962033322 Chr.15: 29137192-29137192 on Build GRCh38
rs777932036 Chr.15: 29137113-29137113 on Build GRCh38
rs751350862 Chr.15: 29137052-29137052 on Build GRCh38
rs778806658 Chr.15: 29137069-29137069 on Build GRCh38
rs768291963 Chr.15: 29137187-29137187 on Build GRCh38
rs1028139505 Chr.15: 29137207-29137207 on Build GRCh38
rs184483466 Chr.15: 29137186-29137186 on Build GRCh38
rs779517068 Chr.15: 29137090-29137090 on Build GRCh38
rs764555440 Chr.15: 29137156-29137156 on Build GRCh38
rs1019217690 Chr.15: 29137191-29137191 on Build GRCh38
rs758043352 Chr.15: 29137023-29137023 on Build GRCh38
rs371324249 Chr.15: 29137021-29137021 on Build GRCh38
rs368449061 Chr.15: 29137118-29137118 on Build GRCh38
rs114821976 Chr.15: 29137197-29137197 on Build GRCh38
rs1046300306 Chr.15: 29137051-29137051 on Build GRCh38
rs549979484 Chr.15: 29137114-29137114 on Build GRCh38
rs868408623 Chr.15: 29137153-29137153 on Build GRCh38
rs553937992 Chr.15: 29137065-29137065 on Build GRCh38
rs774113384 Chr.15: 29137218-29137218 on Build GRCh38
rs138047659 Chr.15: 29137006-29137006 on Build GRCh38
rs556713434 Chr.15: 29137137-29137137 on Build GRCh38
rs1033427997 Chr.15: 29137163-29137163 on Build GRCh38
rs761263422 Chr.15: 29137146-29137146 on Build GRCh38
rs1039183856 Chr.15: 29137038-29137038 on Build GRCh38
rs906174582 Chr.15: 29137079-29137079 on Build GRCh38
rs2292628 Chr.15: 29137143-29137143 on Build GRCh38
rs866429625 Chr.15: 29137169-29137169 on Build GRCh38
rs748122982 Chr.15: 29137097-29137097 on Build GRCh38
rs574184275 Chr.15: 29137070-29137070 on Build GRCh38
rs779722043 Chr.15: 29137047-29137047 on Build GRCh38
rs929325825 Chr.15: 29137020-29137020 on Build GRCh38
rs745845775 Chr.15: 29137022-29137022 on Build GRCh38
rs889308010 Chr.15: 29137183-29137183 on Build GRCh38
rs769723405 Chr.15: 29137110-29137110 on Build GRCh38

View Full Product Details