Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 24958855-24959124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST7
Entrez Gene ID
8530
Gene Name
cystatin F
Gene Aliases
CMAP
UniGene
Hs.143212
Gene Chromosome Location
Chr.20: 24949230-24959928 on Build GRCh38
Transcripts
NM_003650.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1580743 c.347T>C p.I116T Chr.20: 24958965-24958965 on Build GRCh38
6487621 c.330T>C p.Y110Y Chr.20: 24958948-24958948 on Build GRCh38
5757876 c.420G>A p.L140L Chr.20: 24959038-24959038 on Build GRCh38
1580744 c.354A>C p.R118S Chr.20: 24958972-24958972 on Build GRCh38
2887223 c.369A>G p.K123K Chr.20: 24958987-24958987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056033 Chr.20: 24958954-24958954 on Build GRCh38
rs753180769 Chr.20: 24958929-24958929 on Build GRCh38
rs772861443 Chr.20: 24959055-24959055 on Build GRCh38
rs764539062 Chr.20: 24959087-24959087 on Build GRCh38
rs775155557 Chr.20: 24958950-24958950 on Build GRCh38
rs371415469 Chr.20: 24959000-24959000 on Build GRCh38
rs201911475 Chr.20: 24958994-24958994 on Build GRCh38
rs1011155666 Chr.20: 24958898-24958898 on Build GRCh38
rs765936037 Chr.20: 24958921-24958921 on Build GRCh38
rs559940770 Chr.20: 24958906-24958906 on Build GRCh38
rs774205979 Chr.20: 24959030-24959030 on Build GRCh38
rs1021576178 Chr.20: 24958899-24958899 on Build GRCh38
rs770020061 Chr.20: 24959001-24959001 on Build GRCh38
rs368366585 Chr.20: 24958881-24958881 on Build GRCh38
rs544673784 Chr.20: 24958960-24958960 on Build GRCh38
rs768608048 Chr.20: 24959025-24959025 on Build GRCh38
rs757217315 Chr.20: 24958878-24958878 on Build GRCh38
rs1037608243 Chr.20: 24959084-24959084 on Build GRCh38
rs141828767 Chr.20: 24958965-24958965 on Build GRCh38
rs760386858 Chr.20: 24959069-24959069 on Build GRCh38
rs767347572 Chr.20: 24958904-24958904 on Build GRCh38
rs138250783 Chr.20: 24958955-24958955 on Build GRCh38
rs780159527 Chr.20: 24958948-24958948 on Build GRCh38
rs779884917 Chr.20: 24959010-24959010 on Build GRCh38
rs903038946 Chr.20: 24958966-24958966 on Build GRCh38
rs778180806 Chr.20: 24958980-24958980 on Build GRCh38
rs377221759 Chr.20: 24958937-24958937 on Build GRCh38
rs982367494 Chr.20: 24958892-24958893 on Build GRCh38
rs757727120 Chr.20: 24958877-24958877 on Build GRCh38
rs752024220 Chr.20: 24959095-24959095 on Build GRCh38
rs771107712 Chr.20: 24958931-24958931 on Build GRCh38
rs753233539 Chr.20: 24958930-24958930 on Build GRCh38
rs550186726 Chr.20: 24959093-24959093 on Build GRCh38
rs761871362 Chr.20: 24959043-24959043 on Build GRCh38
rs764895498 Chr.20: 24959012-24959013 on Build GRCh38
rs757767902 Chr.20: 24959096-24959096 on Build GRCh38
rs746114061 Chr.20: 24958938-24958938 on Build GRCh38
rs756957641 Chr.20: 24958897-24958897 on Build GRCh38
rs928281229 Chr.20: 24958911-24958911 on Build GRCh38
rs753074395 Chr.20: 24959075-24959075 on Build GRCh38
rs34840829 Chr.20: 24959092-24959093 on Build GRCh38
rs768534697 Chr.20: 24958952-24958952 on Build GRCh38
rs530407849 Chr.20: 24959048-24959048 on Build GRCh38
rs769541436 Chr.20: 24958939-24958939 on Build GRCh38
rs758935314 Chr.20: 24958972-24958972 on Build GRCh38
rs753061639 Chr.20: 24958968-24958968 on Build GRCh38
rs564651753 Chr.20: 24959013-24959013 on Build GRCh38
rs997239221 Chr.20: 24959026-24959028 on Build GRCh38

View Full Product Details