Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48660461-48660702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL2
Entrez Gene ID
54934
Gene Name
KAT8 regulatory NSL complex subunit 2
Gene Aliases
C12orf41, NSL2
UniGene
Hs.505412
Gene Chromosome Location
Chr.12: 48653211-48682252 on Build GRCh38
Transcripts
NM_017822.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3728961 c.974-7delT p.? Chr.12: 48660626-48660626 on Build GRCh38
279255 c.1120G>A p.E374K Chr.12: 48660473-48660473 on Build GRCh38
7372918 c.1079G>T p.C360F Chr.12: 48660514-48660514 on Build GRCh38
7372920 c.1078T>C p.C360R Chr.12: 48660515-48660515 on Build GRCh38
1606183 c.1015_1016insC p.Q339fs*4 Chr.12: 48660577-48660578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762539597 Chr.12: 48660562-48660562 on Build GRCh38
rs775082174 Chr.12: 48660548-48660548 on Build GRCh38
rs748235747 Chr.12: 48660507-48660507 on Build GRCh38
rs781634674 Chr.12: 48660656-48660656 on Build GRCh38
rs369490302 Chr.12: 48660587-48660587 on Build GRCh38
rs1020178544 Chr.12: 48660500-48660500 on Build GRCh38
rs778926782 Chr.12: 48660498-48660498 on Build GRCh38
rs201343803 Chr.12: 48660495-48660495 on Build GRCh38
rs770428448 Chr.12: 48660543-48660543 on Build GRCh38
rs750339885 Chr.12: 48660484-48660484 on Build GRCh38
rs758469276 Chr.12: 48660631-48660631 on Build GRCh38
rs571378936 Chr.12: 48660640-48660641 on Build GRCh38
rs753835313 Chr.12: 48660623-48660623 on Build GRCh38
rs771087847 Chr.12: 48660653-48660653 on Build GRCh38
rs778048394 Chr.12: 48660510-48660510 on Build GRCh38
rs375462172 Chr.12: 48660542-48660542 on Build GRCh38
rs971583045 Chr.12: 48660558-48660558 on Build GRCh38
rs772228059 Chr.12: 48660509-48660509 on Build GRCh38
rs751194055 Chr.12: 48660579-48660579 on Build GRCh38
rs538478628 Chr.12: 48660660-48660660 on Build GRCh38
rs988661862 Chr.12: 48660493-48660493 on Build GRCh38
rs544462966 Chr.12: 48660603-48660603 on Build GRCh38
rs747232555 Chr.12: 48660525-48660525 on Build GRCh38
rs191655655 Chr.12: 48660613-48660613 on Build GRCh38
rs1047116877 Chr.12: 48660635-48660635 on Build GRCh38
rs776137302 Chr.12: 48660664-48660664 on Build GRCh38
rs749853083 Chr.12: 48660666-48660666 on Build GRCh38
rs763578378 Chr.12: 48660569-48660569 on Build GRCh38
rs780890886 Chr.12: 48660483-48660483 on Build GRCh38
rs767054866 Chr.12: 48660529-48660529 on Build GRCh38
rs187783552 Chr.12: 48660651-48660651 on Build GRCh38
rs368928038 Chr.12: 48660659-48660659 on Build GRCh38
rs955316718 Chr.12: 48660501-48660501 on Build GRCh38
rs896725754 Chr.12: 48660638-48660638 on Build GRCh38
rs183564434 Chr.12: 48660650-48660650 on Build GRCh38
rs767295406 Chr.12: 48660596-48660596 on Build GRCh38
rs372786876 Chr.12: 48660604-48660604 on Build GRCh38
rs986641476 Chr.12: 48660504-48660504 on Build GRCh38
rs776878662 Chr.12: 48660536-48660536 on Build GRCh38

View Full Product Details