Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38880859-38881132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT2
Entrez Gene ID
22933
Gene Name
sirtuin 2
Gene Aliases
SIR2, SIR2L, SIR2L2
UniGene
Hs.466693
Gene Chromosome Location
Chr.19: 38878555-38899862 on Build GRCh38
Transcripts
NM_001193286.1 NM_012237.3 NM_030593.2 NR_034146.1 XM_006723111.1 XM_011526654.1 XM_011526655.1 XM_017026500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180424 c.718C>A p.P240T Chr.19: 38881129-38881129 on Build GRCh38
6778829 c.782G>A p.G261D Chr.19: 38880863-38880863 on Build GRCh38
6041266 c.723G>A p.A241A Chr.19: 38881124-38881124 on Build GRCh38
5800026 c.738T>A p.C246* Chr.19: 38881109-38881109 on Build GRCh38
5972926 c.768C>G p.L256L Chr.19: 38880877-38880877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200181575 Chr.19: 38880920-38880920 on Build GRCh38
rs564687367 Chr.19: 38880904-38880904 on Build GRCh38
rs201755577 Chr.19: 38880916-38880916 on Build GRCh38
rs372301365 Chr.19: 38880930-38880930 on Build GRCh38
rs200919229 Chr.19: 38880912-38880912 on Build GRCh38
rs202222176 Chr.19: 38880941-38880941 on Build GRCh38
rs112437333 Chr.19: 38881067-38881067 on Build GRCh38
rs183190025 Chr.19: 38880929-38880929 on Build GRCh38
rs747546861 Chr.19: 38880907-38880907 on Build GRCh38
rs753566297 Chr.19: 38881051-38881051 on Build GRCh38
rs374379865 Chr.19: 38880906-38880906 on Build GRCh38
rs892168700 Chr.19: 38880954-38880954 on Build GRCh38
rs537367346 Chr.19: 38881006-38881006 on Build GRCh38
rs201896797 Chr.19: 38880902-38880902 on Build GRCh38
rs761852958 Chr.19: 38880939-38880939 on Build GRCh38
rs200078767 Chr.19: 38880908-38880908 on Build GRCh38
rs1003028481 Chr.19: 38880998-38880998 on Build GRCh38
rs774573551 Chr.19: 38881080-38881080 on Build GRCh38
rs535569038 Chr.19: 38880947-38880947 on Build GRCh38
rs891877229 Chr.19: 38881045-38881045 on Build GRCh38
rs558298927 Chr.19: 38880984-38880985 on Build GRCh38
rs755831873 Chr.19: 38880891-38880891 on Build GRCh38
rs1048539766 Chr.19: 38880961-38880961 on Build GRCh38
rs963319254 Chr.19: 38881008-38881008 on Build GRCh38
rs748304191 Chr.19: 38881083-38881083 on Build GRCh38
rs768700376 Chr.19: 38881077-38881077 on Build GRCh38
rs773330777 Chr.19: 38881091-38881091 on Build GRCh38
rs746365489 Chr.19: 38881066-38881066 on Build GRCh38
rs985261189 Chr.19: 38880917-38880917 on Build GRCh38
rs1030003106 Chr.19: 38880959-38880959 on Build GRCh38
rs113433881 Chr.19: 38880982-38880982 on Build GRCh38
rs767492808 Chr.19: 38880940-38880940 on Build GRCh38
rs763067102 Chr.19: 38880921-38880921 on Build GRCh38
rs375607212 Chr.19: 38881092-38881092 on Build GRCh38
rs201430878 Chr.19: 38881061-38881061 on Build GRCh38
rs766228345 Chr.19: 38881093-38881093 on Build GRCh38
rs146544643 Chr.19: 38881065-38881065 on Build GRCh38
rs919178953 Chr.19: 38881044-38881044 on Build GRCh38
rs780236940 Chr.19: 38881071-38881071 on Build GRCh38
rs909434860 Chr.19: 38880974-38880974 on Build GRCh38
rs779821357 Chr.19: 38880901-38880901 on Build GRCh38
rs973357494 Chr.19: 38881023-38881023 on Build GRCh38
rs113989943 Chr.19: 38881076-38881076 on Build GRCh38
rs550989255 Chr.19: 38880983-38880983 on Build GRCh38
rs770141173 Chr.19: 38880914-38880914 on Build GRCh38
rs201927970 Chr.19: 38881110-38881110 on Build GRCh38
rs879425449 Chr.19: 38880924-38880924 on Build GRCh38
rs776239943 Chr.19: 38881104-38881104 on Build GRCh38
rs199524470 Chr.19: 38881062-38881062 on Build GRCh38
rs200401706 Chr.19: 38880946-38880946 on Build GRCh38
rs569128269 Chr.19: 38880984-38880984 on Build GRCh38
rs985957590 Chr.19: 38880989-38880989 on Build GRCh38
rs201596933 Chr.19: 38881084-38881084 on Build GRCh38
rs112848414 Chr.19: 38880911-38880911 on Build GRCh38
rs1009190611 Chr.19: 38880956-38880956 on Build GRCh38
rs200562565 Chr.19: 38881064-38881064 on Build GRCh38
rs983778630 Chr.19: 38881106-38881106 on Build GRCh38
rs905423586 Chr.19: 38880938-38880938 on Build GRCh38

View Full Product Details