Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93634727-93634958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6C
Entrez Gene ID
5146
Gene Name
phosphodiesterase 6C
Gene Aliases
ACHM5, COD4, PDEA2
UniGene
Hs.658121
Gene Chromosome Location
Chr.10: 93612588-93665673 on Build GRCh38
Transcripts
NM_006204.3 XM_017016322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5371328 c.1171C>T p.P391S Chr.10: 93634809-93634809 on Build GRCh38
6067228 c.1123G>T p.G375* Chr.10: 93634761-93634761 on Build GRCh38
921596 c.1189G>T p.E397* Chr.10: 93634827-93634827 on Build GRCh38
3441704 c.1135G>A p.E379K Chr.10: 93634773-93634773 on Build GRCh38
3441705 c.1168C>T p.L390L Chr.10: 93634806-93634806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768524833 Chr.10: 93634781-93634781 on Build GRCh38
rs997479152 Chr.10: 93634892-93634892 on Build GRCh38
rs148611776 Chr.10: 93634773-93634773 on Build GRCh38
rs200771894 Chr.10: 93634772-93634772 on Build GRCh38
rs746820022 Chr.10: 93634888-93634888 on Build GRCh38
rs755562979 Chr.10: 93634779-93634779 on Build GRCh38
rs1031796960 Chr.10: 93634923-93634923 on Build GRCh38
rs776602508 Chr.10: 93634788-93634788 on Build GRCh38
rs750495412 Chr.10: 93634872-93634872 on Build GRCh38
rs764573887 Chr.10: 93634850-93634850 on Build GRCh38
rs761000143 Chr.10: 93634925-93634925 on Build GRCh38
rs772639930 Chr.10: 93634810-93634810 on Build GRCh38
rs753049268 Chr.10: 93634844-93634844 on Build GRCh38
rs758720479 Chr.10: 93634880-93634880 on Build GRCh38
rs199948964 Chr.10: 93634849-93634849 on Build GRCh38
rs757784780 Chr.10: 93634851-93634851 on Build GRCh38
rs779140836 Chr.10: 93634861-93634861 on Build GRCh38
rs761298393 Chr.10: 93634793-93634793 on Build GRCh38
rs371276239 Chr.10: 93634917-93634917 on Build GRCh38
rs762424340 Chr.10: 93634919-93634919 on Build GRCh38
rs768495549 Chr.10: 93634889-93634889 on Build GRCh38
rs779339015 Chr.10: 93634764-93634764 on Build GRCh38
rs765966527 Chr.10: 93634814-93634814 on Build GRCh38
rs530593118 Chr.10: 93634904-93634904 on Build GRCh38
rs764534931 Chr.10: 93634926-93634926 on Build GRCh38
rs202050878 Chr.10: 93634899-93634899 on Build GRCh38
rs866611156 Chr.10: 93634866-93634866 on Build GRCh38
rs772358173 Chr.10: 93634768-93634768 on Build GRCh38
rs141384587 Chr.10: 93634812-93634812 on Build GRCh38
rs201268183 Chr.10: 93634929-93634929 on Build GRCh38
rs780401922 Chr.10: 93634881-93634881 on Build GRCh38
rs193276411 Chr.10: 93634924-93634924 on Build GRCh38
rs200108487 Chr.10: 93634804-93634804 on Build GRCh38
rs746108727 Chr.10: 93634767-93634767 on Build GRCh38
rs998950625 Chr.10: 93634914-93634914 on Build GRCh38
rs144591862 Chr.10: 93634902-93634902 on Build GRCh38
rs772729811 Chr.10: 93634912-93634912 on Build GRCh38
rs879067314 Chr.10: 93634777-93634777 on Build GRCh38

View Full Product Details