Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 161227164-161227425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMD3
Entrez Gene ID
51068
Gene Name
NMD3 ribosome export adaptor
Gene Aliases
CGI-07
UniGene
Hs.598836
Gene Chromosome Location
Chr.3: 161221262-161253532 on Build GRCh38
Transcripts
NM_001320227.1 NM_015938.4 XM_005247511.1 XM_005247512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7232102 c.187C>T p.Q63* Chr.3: 161227254-161227254 on Build GRCh38
1327747 c.197G>T p.G66V Chr.3: 161227264-161227264 on Build GRCh38
4114932 c.249G>C p.L83F Chr.3: 161227316-161227316 on Build GRCh38
3202414 c.263C>T p.A88V Chr.3: 161227330-161227330 on Build GRCh38
1217511 c.202T>C p.W68R Chr.3: 161227269-161227269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145565492 Chr.3: 161227379-161227379 on Build GRCh38
rs776141363 Chr.3: 161227314-161227314 on Build GRCh38
rs376527627 Chr.3: 161227330-161227330 on Build GRCh38
rs781086065 Chr.3: 161227262-161227262 on Build GRCh38
rs770438046 Chr.3: 161227394-161227394 on Build GRCh38
rs949653376 Chr.3: 161227320-161227320 on Build GRCh38
rs752772857 Chr.3: 161227214-161227214 on Build GRCh38
rs1008794637 Chr.3: 161227284-161227284 on Build GRCh38
rs200969579 Chr.3: 161227256-161227256 on Build GRCh38
rs755633966 Chr.3: 161227381-161227381 on Build GRCh38
rs150533598 Chr.3: 161227266-161227266 on Build GRCh38
rs774536225 Chr.3: 161227304-161227304 on Build GRCh38
rs751599334 Chr.3: 161227372-161227372 on Build GRCh38
rs374116167 Chr.3: 161227315-161227315 on Build GRCh38
rs746999503 Chr.3: 161227253-161227253 on Build GRCh38
rs747695994 Chr.3: 161227265-161227265 on Build GRCh38
rs754103534 Chr.3: 161227228-161227228 on Build GRCh38
rs748999631 Chr.3: 161227392-161227392 on Build GRCh38
rs777196677 Chr.3: 161227382-161227382 on Build GRCh38
rs759890891 Chr.3: 161227212-161227212 on Build GRCh38
rs932784961 Chr.3: 161227197-161227197 on Build GRCh38
rs764481307 Chr.3: 161227226-161227226 on Build GRCh38
rs371100073 Chr.3: 161227222-161227222 on Build GRCh38
rs748833148 Chr.3: 161227274-161227274 on Build GRCh38
rs138466882 Chr.3: 161227308-161227308 on Build GRCh38
rs941730427 Chr.3: 161227385-161227385 on Build GRCh38
rs370575163 Chr.3: 161227331-161227331 on Build GRCh38
rs181252398 Chr.3: 161227357-161227357 on Build GRCh38
rs763855439 Chr.3: 161227393-161227394 on Build GRCh38
rs760108317 Chr.3: 161227306-161227306 on Build GRCh38
rs139379354 Chr.3: 161227250-161227250 on Build GRCh38
rs764161771 Chr.3: 161227363-161227363 on Build GRCh38
rs781547354 Chr.3: 161227276-161227276 on Build GRCh38
rs888975292 Chr.3: 161227260-161227260 on Build GRCh38
rs771329468 Chr.3: 161227299-161227299 on Build GRCh38
rs755353137 Chr.3: 161227272-161227272 on Build GRCh38
rs201509848 Chr.3: 161227229-161227229 on Build GRCh38
rs764215323 Chr.3: 161227316-161227316 on Build GRCh38

View Full Product Details