Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31262049-31262322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIMK2
Entrez Gene ID
3985
Gene Name
LIM domain kinase 2
Gene Aliases
-
UniGene
Hs.474596
Gene Chromosome Location
Chr.22: 31212264-31280080 on Build GRCh38
Transcripts
NM_001031801.1 NM_005569.3 NM_016733.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6332392 c.502C>A p.H168N Chr.22: 31262147-31262147 on Build GRCh38
1242464 c.559A>T p.N187Y Chr.22: 31262204-31262204 on Build GRCh38
4582329 c.607C>T p.R203C Chr.22: 31262189-31262189 on Build GRCh38
1256492 c.545G>A p.R182H Chr.22: 31262190-31262190 on Build GRCh38
6701300 c.559C>T p.R187W Chr.22: 31262141-31262141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761645334 Chr.22: 31262279-31262279 on Build GRCh38
rs115736350 Chr.22: 31262234-31262234 on Build GRCh38
rs781151105 Chr.22: 31262215-31262215 on Build GRCh38
rs772419303 Chr.22: 31262115-31262115 on Build GRCh38
rs777185924 Chr.22: 31262236-31262236 on Build GRCh38
rs2073857 Chr.22: 31262295-31262295 on Build GRCh38
rs757149319 Chr.22: 31262205-31262205 on Build GRCh38
rs775183685 Chr.22: 31262084-31262084 on Build GRCh38
rs577293364 Chr.22: 31262143-31262143 on Build GRCh38
rs200700695 Chr.22: 31262252-31262252 on Build GRCh38
rs371699959 Chr.22: 31262171-31262171 on Build GRCh38
rs751350349 Chr.22: 31262190-31262190 on Build GRCh38
rs5997927 Chr.22: 31262170-31262170 on Build GRCh38
rs115204987 Chr.22: 31262229-31262229 on Build GRCh38
rs376569162 Chr.22: 31262214-31262214 on Build GRCh38
rs200565424 Chr.22: 31262172-31262172 on Build GRCh38
rs768236818 Chr.22: 31262228-31262228 on Build GRCh38
rs773495668 Chr.22: 31262141-31262141 on Build GRCh38
rs770340281 Chr.22: 31262245-31262245 on Build GRCh38
rs748996101 Chr.22: 31262089-31262089 on Build GRCh38
rs750367200 Chr.22: 31262291-31262291 on Build GRCh38
rs767272954 Chr.22: 31262280-31262280 on Build GRCh38
rs760029773 Chr.22: 31262237-31262237 on Build GRCh38
rs752962006 Chr.22: 31262177-31262177 on Build GRCh38
rs747348171 Chr.22: 31262230-31262230 on Build GRCh38
rs761037866 Chr.22: 31262142-31262142 on Build GRCh38
rs939100847 Chr.22: 31262090-31262090 on Build GRCh38
rs779494391 Chr.22: 31262220-31262220 on Build GRCh38
rs766874005 Chr.22: 31262158-31262158 on Build GRCh38
rs34930775 Chr.22: 31262219-31262219 on Build GRCh38
rs369493707 Chr.22: 31262216-31262216 on Build GRCh38
rs751869792 Chr.22: 31262260-31262260 on Build GRCh38
rs200312431 Chr.22: 31262189-31262189 on Build GRCh38
rs764370739 Chr.22: 31262259-31262259 on Build GRCh38

View Full Product Details