Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2317191-2317416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGM3
Entrez Gene ID
7053
Gene Name
transglutaminase 3
Gene Aliases
TGE
UniGene
Hs.2022
Gene Chromosome Location
Chr.20: 2295967-2341079 on Build GRCh38
Transcripts
NM_003245.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
443538 c.884C>A p.T295N Chr.20: 2317386-2317386 on Build GRCh38
1410739 c.806G>A p.R269Q Chr.20: 2317204-2317204 on Build GRCh38
5916855 c.872C>T p.S291F Chr.20: 2317374-2317374 on Build GRCh38
2705947 c.827T>C p.F276S Chr.20: 2317225-2317225 on Build GRCh38
3785356 c.836C>A p.T279N Chr.20: 2317234-2317234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763093832 Chr.20: 2317281-2317281 on Build GRCh38
rs372303255 Chr.20: 2317249-2317249 on Build GRCh38
rs773530017 Chr.20: 2317332-2317332 on Build GRCh38
rs763351550 Chr.20: 2317219-2317219 on Build GRCh38
rs145397721 Chr.20: 2317376-2317376 on Build GRCh38
rs750569681 Chr.20: 2317259-2317259 on Build GRCh38
rs756238647 Chr.20: 2317262-2317262 on Build GRCh38
rs6137668 Chr.20: 2317279-2317279 on Build GRCh38
rs369240674 Chr.20: 2317257-2317257 on Build GRCh38
rs774929160 Chr.20: 2317327-2317327 on Build GRCh38
rs965635840 Chr.20: 2317349-2317349 on Build GRCh38
rs750134199 Chr.20: 2317372-2317372 on Build GRCh38
rs146568830 Chr.20: 2317377-2317377 on Build GRCh38
rs772526693 Chr.20: 2317395-2317395 on Build GRCh38
rs756581828 Chr.20: 2317331-2317331 on Build GRCh38
rs776091527 Chr.20: 2317210-2317210 on Build GRCh38
rs1055871365 Chr.20: 2317388-2317388 on Build GRCh38
rs748641552 Chr.20: 2317390-2317390 on Build GRCh38
rs201000537 Chr.20: 2317351-2317351 on Build GRCh38
rs201739042 Chr.20: 2317341-2317341 on Build GRCh38
rs200046805 Chr.20: 2317310-2317310 on Build GRCh38
rs200047268 Chr.20: 2317355-2317355 on Build GRCh38
rs549450324 Chr.20: 2317256-2317256 on Build GRCh38
rs759556192 Chr.20: 2317343-2317343 on Build GRCh38
rs780932147 Chr.20: 2317309-2317309 on Build GRCh38
rs866615963 Chr.20: 2317350-2317350 on Build GRCh38
rs376434552 Chr.20: 2317334-2317334 on Build GRCh38
rs768237602 Chr.20: 2317389-2317389 on Build GRCh38
rs769295727 Chr.20: 2317321-2317321 on Build GRCh38
rs757069005 Chr.20: 2317356-2317356 on Build GRCh38
rs749864159 Chr.20: 2317319-2317319 on Build GRCh38
rs777602982 Chr.20: 2317291-2317291 on Build GRCh38
rs764616112 Chr.20: 2317227-2317227 on Build GRCh38
rs753848915 Chr.20: 2317338-2317338 on Build GRCh38
rs766347571 Chr.20: 2317271-2317271 on Build GRCh38
rs774725338 Chr.20: 2317379-2317379 on Build GRCh38
rs369661811 Chr.20: 2317335-2317335 on Build GRCh38
rs762208603 Chr.20: 2317253-2317253 on Build GRCh38
rs756981026 Chr.20: 2317304-2317304 on Build GRCh38
rs751371284 Chr.20: 2317293-2317293 on Build GRCh38
rs780831622 Chr.20: 2317360-2317360 on Build GRCh38
rs557107089 Chr.20: 2317298-2317298 on Build GRCh38
rs758328163 Chr.20: 2317286-2317286 on Build GRCh38
rs369015761 Chr.20: 2317284-2317284 on Build GRCh38

View Full Product Details