Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60842803-60843030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC86
Entrez Gene ID
79080
Gene Name
coiled-coil domain containing 86
Gene Aliases
-
UniGene
Hs.4253
Gene Chromosome Location
Chr.11: 60841956-60851088 on Build GRCh38
Transcripts
NM_024098.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5430284 c.742G>C p.D248H Chr.11: 60842866-60842866 on Build GRCh38
415518 c.708G>A p.P236P Chr.11: 60842832-60842832 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs566323886 Chr.11: 60842947-60842947 on Build GRCh38
rs769939589 Chr.11: 60842894-60842894 on Build GRCh38
rs755676252 Chr.11: 60842932-60842932 on Build GRCh38
rs776948950 Chr.11: 60842890-60842890 on Build GRCh38
rs752447117 Chr.11: 60842930-60842930 on Build GRCh38
rs1039744296 Chr.11: 60842980-60842980 on Build GRCh38
rs1045666144 Chr.11: 60842901-60842901 on Build GRCh38
rs773125938 Chr.11: 60842909-60842909 on Build GRCh38
rs535181700 Chr.11: 60842975-60842975 on Build GRCh38
rs934500807 Chr.11: 60842937-60842937 on Build GRCh38
rs746412311 Chr.11: 60842865-60842865 on Build GRCh38
rs917497883 Chr.11: 60842929-60842929 on Build GRCh38
rs962751262 Chr.11: 60842842-60842842 on Build GRCh38
rs752289054 Chr.11: 60842832-60842832 on Build GRCh38
rs1052873294 Chr.11: 60842990-60842990 on Build GRCh38
rs773043105 Chr.11: 60842886-60842886 on Build GRCh38
rs980477641 Chr.11: 60842854-60842854 on Build GRCh38
rs946245115 Chr.11: 60842938-60842938 on Build GRCh38
rs774419887 Chr.11: 60842927-60842927 on Build GRCh38
rs758865357 Chr.11: 60842866-60842866 on Build GRCh38
rs748291836 Chr.11: 60842893-60842893 on Build GRCh38
rs777129864 Chr.11: 60842838-60842838 on Build GRCh38
rs762987069 Chr.11: 60842910-60842910 on Build GRCh38
rs759472767 Chr.11: 60842928-60842928 on Build GRCh38
rs192692508 Chr.11: 60842995-60842995 on Build GRCh38
rs926078849 Chr.11: 60842949-60842949 on Build GRCh38
rs936111249 Chr.11: 60842968-60842968 on Build GRCh38
rs552381645 Chr.11: 60842942-60842942 on Build GRCh38
rs780403526 Chr.11: 60842870-60842870 on Build GRCh38
rs766360300 Chr.11: 60842924-60842924 on Build GRCh38

View Full Product Details