Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39509535-39509808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGDH
Entrez Gene ID
7358
Gene Name
UDP-glucose 6-dehydrogenase
Gene Aliases
GDH, UDP-GlcDH, UDPGDH, UGD
UniGene
Hs.572518
Gene Chromosome Location
Chr.4: 39498755-39527598 on Build GRCh38
Transcripts
NM_001184700.1 NM_001184701.1 NM_003359.3 XM_005262667.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910350 c.793T>C p.F265L Chr.4: 39509778-39509778 on Build GRCh38
733508 c.777G>T p.Q259H Chr.4: 39509794-39509794 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999942279 Chr.4: 39509583-39509583 on Build GRCh38
rs942357391 Chr.4: 39509635-39509635 on Build GRCh38
rs556478323 Chr.4: 39509686-39509686 on Build GRCh38
rs894126186 Chr.4: 39509649-39509649 on Build GRCh38
rs1044007255 Chr.4: 39509564-39509564 on Build GRCh38
rs73137495 Chr.4: 39509680-39509680 on Build GRCh38
rs746355905 Chr.4: 39509729-39509729 on Build GRCh38
rs925601917 Chr.4: 39509592-39509592 on Build GRCh38
rs778306943 Chr.4: 39509713-39509713 on Build GRCh38
rs747339465 Chr.4: 39509726-39509726 on Build GRCh38
rs116145557 Chr.4: 39509779-39509779 on Build GRCh38
rs982252575 Chr.4: 39509721-39509721 on Build GRCh38
rs1023906753 Chr.4: 39509682-39509682 on Build GRCh38
rs774522828 Chr.4: 39509755-39509755 on Build GRCh38
rs775372571 Chr.4: 39509736-39509736 on Build GRCh38
rs971348318 Chr.4: 39509685-39509685 on Build GRCh38
rs766146242 Chr.4: 39509769-39509769 on Build GRCh38
rs6531722 Chr.4: 39509739-39509739 on Build GRCh38
rs1032701129 Chr.4: 39509633-39509633 on Build GRCh38
rs760508476 Chr.4: 39509765-39509765 on Build GRCh38
rs777740224 Chr.4: 39509697-39509697 on Build GRCh38
rs768823934 Chr.4: 39509742-39509742 on Build GRCh38
rs560741608 Chr.4: 39509608-39509608 on Build GRCh38
rs769921309 Chr.4: 39509734-39509734 on Build GRCh38
rs57073600 Chr.4: 39509775-39509775 on Build GRCh38
rs997965352 Chr.4: 39509706-39509706 on Build GRCh38
rs201553424 Chr.4: 39509770-39509770 on Build GRCh38
rs886950933 Chr.4: 39509768-39509768 on Build GRCh38
rs371038993 Chr.4: 39509728-39509728 on Build GRCh38
rs377571623 Chr.4: 39509582-39509583 on Build GRCh38
rs540936674 Chr.4: 39509578-39509578 on Build GRCh38
rs376382481 Chr.4: 39509727-39509727 on Build GRCh38

View Full Product Details