Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8303810-8304079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD320
Entrez Gene ID
51293
Gene Name
CD320 molecule
Gene Aliases
8D6, 8D6A, TCBLR
UniGene
Hs.558499
Gene Chromosome Location
Chr.19: 8302127-8308356 on Build GRCh38
Transcripts
NM_001165895.1 NM_016579.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1397748 c.386G>A p.R129H Chr.19: 8303971-8303971 on Build GRCh38
4083017 c.451C>T p.R151C Chr.19: 8303906-8303906 on Build GRCh38
4749580 c.419C>T p.T140M Chr.19: 8303938-8303938 on Build GRCh38
4128002 c.295C>T p.Q99* Chr.19: 8304062-8304062 on Build GRCh38
4987615 c.374G>A p.R125H Chr.19: 8303983-8303983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772544276 Chr.19: 8303861-8303861 on Build GRCh38
rs188158114 Chr.19: 8303879-8303879 on Build GRCh38
rs767324816 Chr.19: 8303930-8303930 on Build GRCh38
rs559766397 Chr.19: 8304040-8304040 on Build GRCh38
rs146218039 Chr.19: 8303919-8303919 on Build GRCh38
rs1044641805 Chr.19: 8304050-8304050 on Build GRCh38
rs895250025 Chr.19: 8304047-8304047 on Build GRCh38
rs745552673 Chr.19: 8303993-8303993 on Build GRCh38
rs762418586 Chr.19: 8303917-8303917 on Build GRCh38
rs755200274 Chr.19: 8303960-8303960 on Build GRCh38
rs777346987 Chr.19: 8303842-8303842 on Build GRCh38
rs144097701 Chr.19: 8303979-8303979 on Build GRCh38
rs139064611 Chr.19: 8303971-8303971 on Build GRCh38
rs866153489 Chr.19: 8304014-8304014 on Build GRCh38
rs760432859 Chr.19: 8303932-8303932 on Build GRCh38
rs2232784 Chr.19: 8303874-8303874 on Build GRCh38
rs758476844 Chr.19: 8303951-8303951 on Build GRCh38
rs886297543 Chr.19: 8303986-8303986 on Build GRCh38
rs755073624 Chr.19: 8304051-8304051 on Build GRCh38
rs751841592 Chr.19: 8303953-8303953 on Build GRCh38
rs747252781 Chr.19: 8303905-8303905 on Build GRCh38
rs201828443 Chr.19: 8303880-8303880 on Build GRCh38
rs780202188 Chr.19: 8303903-8303903 on Build GRCh38
rs201166605 Chr.19: 8303945-8303945 on Build GRCh38
rs369646365 Chr.19: 8304039-8304039 on Build GRCh38
rs201624090 Chr.19: 8303911-8303911 on Build GRCh38
rs761755805 Chr.19: 8304038-8304038 on Build GRCh38
rs762341463 Chr.19: 8303854-8303854 on Build GRCh38
rs540057476 Chr.19: 8304024-8304024 on Build GRCh38
rs773734293 Chr.19: 8303914-8303914 on Build GRCh38
rs776439222 Chr.19: 8304027-8304027 on Build GRCh38
rs760524953 Chr.19: 8304020-8304020 on Build GRCh38
rs948576208 Chr.19: 8304023-8304023 on Build GRCh38
rs2232783 Chr.19: 8303910-8303910 on Build GRCh38
rs1003366693 Chr.19: 8304001-8304001 on Build GRCh38
rs777691789 Chr.19: 8304024-8304029 on Build GRCh38
rs773940394 Chr.19: 8303923-8303923 on Build GRCh38
rs749795604 Chr.19: 8303972-8303972 on Build GRCh38
rs775103018 Chr.19: 8304017-8304017 on Build GRCh38
rs750510195 Chr.19: 8303898-8303898 on Build GRCh38
rs771751779 Chr.19: 8303995-8303995 on Build GRCh38
rs538142552 Chr.19: 8303969-8303969 on Build GRCh38
rs751202127 Chr.19: 8303867-8303867 on Build GRCh38
rs199545496 Chr.19: 8303862-8303862 on Build GRCh38
rs141674860 Chr.19: 8303912-8303912 on Build GRCh38
rs763991012 Chr.19: 8303944-8303944 on Build GRCh38
rs759382341 Chr.19: 8303868-8303868 on Build GRCh38
rs372005633 Chr.19: 8303952-8303952 on Build GRCh38
rs759132877 Chr.19: 8303929-8303929 on Build GRCh38
rs769438880 Chr.19: 8303860-8303860 on Build GRCh38
rs908196413 Chr.19: 8304003-8304003 on Build GRCh38
rs867215292 Chr.19: 8304043-8304043 on Build GRCh38
rs767294120 Chr.19: 8303870-8303870 on Build GRCh38
rs375310833 Chr.19: 8303983-8303983 on Build GRCh38
rs758093694 Chr.19: 8303969-8303969 on Build GRCh38
rs758485696 Chr.19: 8303901-8303901 on Build GRCh38
rs939686446 Chr.19: 8304013-8304013 on Build GRCh38
rs769150463 Chr.19: 8303906-8303906 on Build GRCh38
rs765987551 Chr.19: 8303864-8303864 on Build GRCh38
rs767897892 Chr.19: 8304052-8304052 on Build GRCh38
rs776558154 Chr.19: 8304021-8304021 on Build GRCh38

View Full Product Details