Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52239362-52239633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE9
Entrez Gene ID
9372
Gene Name
zinc finger FYVE-type containing 9
Gene Aliases
MADHIP, NSP, PPP1R173, SARA, SMADIP
UniGene
Hs.532345
Gene Chromosome Location
Chr.1: 52142001-52348664 on Build GRCh38
Transcripts
NM_004799.3 NM_007324.3 XM_011542437.2 XM_017002845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2192766 c.2035G>T p.G679* Chr.1: 52239452-52239452 on Build GRCh38
7165918 c.2071G>A p.E691K Chr.1: 52239488-52239488 on Build GRCh38
3490653 c.2006C>T p.P669L Chr.1: 52239423-52239423 on Build GRCh38
5014109 c.2036G>T p.G679V Chr.1: 52239453-52239453 on Build GRCh38
3490652 c.1970C>T p.S657F Chr.1: 52239387-52239387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146344711 Chr.1: 52239510-52239510 on Build GRCh38
rs1064884 Chr.1: 52239481-52239481 on Build GRCh38
rs760692476 Chr.1: 52239500-52239500 on Build GRCh38
rs200725080 Chr.1: 52239416-52239416 on Build GRCh38
rs779545853 Chr.1: 52239461-52239461 on Build GRCh38
rs142234365 Chr.1: 52239463-52239463 on Build GRCh38
rs996509057 Chr.1: 52239555-52239555 on Build GRCh38
rs34437486 Chr.1: 52239502-52239502 on Build GRCh38
rs765332808 Chr.1: 52239529-52239529 on Build GRCh38
rs753133737 Chr.1: 52239411-52239411 on Build GRCh38
rs764149475 Chr.1: 52239613-52239614 on Build GRCh38
rs745900026 Chr.1: 52239479-52239479 on Build GRCh38
rs752914815 Chr.1: 52239548-52239548 on Build GRCh38
rs753986747 Chr.1: 52239511-52239511 on Build GRCh38
rs758733784 Chr.1: 52239417-52239417 on Build GRCh38
rs771426652 Chr.1: 52239459-52239459 on Build GRCh38
rs773836104 Chr.1: 52239469-52239469 on Build GRCh38
rs761504416 Chr.1: 52239470-52239470 on Build GRCh38
rs888036275 Chr.1: 52239400-52239400 on Build GRCh38
rs113262686 Chr.1: 52239571-52239571 on Build GRCh38
rs747575639 Chr.1: 52239458-52239458 on Build GRCh38
rs748828816 Chr.1: 52239605-52239605 on Build GRCh38
rs751856296 Chr.1: 52239561-52239561 on Build GRCh38
rs202212629 Chr.1: 52239443-52239443 on Build GRCh38
rs758635376 Chr.1: 52239559-52239559 on Build GRCh38
rs902214159 Chr.1: 52239547-52239547 on Build GRCh38
rs3790523 Chr.1: 52239613-52239613 on Build GRCh38
rs773135717 Chr.1: 52239495-52239495 on Build GRCh38
rs376818411 Chr.1: 52239455-52239455 on Build GRCh38
rs922679050 Chr.1: 52239434-52239434 on Build GRCh38
rs112398067 Chr.1: 52239454-52239454 on Build GRCh38
rs759490228 Chr.1: 52239526-52239526 on Build GRCh38
rs781628187 Chr.1: 52239577-52239577 on Build GRCh38
rs778949561 Chr.1: 52239390-52239390 on Build GRCh38
rs1039672708 Chr.1: 52239399-52239399 on Build GRCh38
rs771819806 Chr.1: 52239476-52239476 on Build GRCh38

View Full Product Details