Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132727343-132727612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK8
Entrez Gene ID
158067
Gene Name
adenylate kinase 8
Gene Aliases
AK 8, C9orf98, DDX31
UniGene
Hs.421340
Gene Chromosome Location
Chr.9: 132725574-132878811 on Build GRCh38
Transcripts
NM_001317958.1 NM_001317959.1 NM_152572.2 XM_005272169.2 XM_006716965.2 XM_011518278.2 XM_017014308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3432954 c.1147G>T p.D383Y Chr.9: 132727509-132727509 on Build GRCh38
5732999 c.1122-1G>C p.? Chr.9: 132727535-132727535 on Build GRCh38
6114942 c.1164G>A p.R388R Chr.9: 132727492-132727492 on Build GRCh38
1624669 c.1202G>T p.R401M Chr.9: 132727454-132727454 on Build GRCh38
6456492 c.1130T>C p.F377S Chr.9: 132727526-132727526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770435981 Chr.9: 132727475-132727475 on Build GRCh38
rs750651827 Chr.9: 132727515-132727515 on Build GRCh38
rs143990720 Chr.9: 132727460-132727460 on Build GRCh38
rs781626145 Chr.9: 132727478-132727478 on Build GRCh38
rs757949836 Chr.9: 132727440-132727440 on Build GRCh38
rs750971727 Chr.9: 132727562-132727562 on Build GRCh38
rs140784839 Chr.9: 132727503-132727503 on Build GRCh38
rs368966104 Chr.9: 132727492-132727492 on Build GRCh38
rs930081394 Chr.9: 132727500-132727500 on Build GRCh38
rs374821225 Chr.9: 132727431-132727431 on Build GRCh38
rs766218182 Chr.9: 132727425-132727425 on Build GRCh38
rs971430840 Chr.9: 132727406-132727406 on Build GRCh38
rs763524400 Chr.9: 132727494-132727494 on Build GRCh38
rs753517737 Chr.9: 132727426-132727426 on Build GRCh38
rs538896732 Chr.9: 132727378-132727378 on Build GRCh38
rs371252329 Chr.9: 132727498-132727498 on Build GRCh38
rs369191618 Chr.9: 132727551-132727551 on Build GRCh38
rs748062104 Chr.9: 132727433-132727433 on Build GRCh38
rs759304341 Chr.9: 132727520-132727520 on Build GRCh38
rs145583710 Chr.9: 132727514-132727514 on Build GRCh38
rs570204054 Chr.9: 132727499-132727499 on Build GRCh38
rs955195115 Chr.9: 132727374-132727374 on Build GRCh38
rs752668275 Chr.9: 132727537-132727537 on Build GRCh38
rs1055770540 Chr.9: 132727531-132727531 on Build GRCh38
rs34599683 Chr.9: 132727551-132727552 on Build GRCh38
rs567355720 Chr.9: 132727546-132727546 on Build GRCh38
rs146410308 Chr.9: 132727493-132727493 on Build GRCh38
rs746288370 Chr.9: 132727480-132727480 on Build GRCh38
rs778887473 Chr.9: 132727432-132727432 on Build GRCh38
rs150108839 Chr.9: 132727513-132727513 on Build GRCh38
rs773893315 Chr.9: 132727439-132727439 on Build GRCh38
rs139630914 Chr.9: 132727522-132727522 on Build GRCh38
rs376208055 Chr.9: 132727511-132727511 on Build GRCh38
rs981520343 Chr.9: 132727421-132727421 on Build GRCh38
rs756789721 Chr.9: 132727564-132727564 on Build GRCh38
rs546096976 Chr.9: 132727443-132727443 on Build GRCh38
rs889158046 Chr.9: 132727372-132727410 on Build GRCh38
rs1050795688 Chr.9: 132727527-132727527 on Build GRCh38
rs962931063 Chr.9: 132727434-132727434 on Build GRCh38
rs780768256 Chr.9: 132727577-132727577 on Build GRCh38
rs746702160 Chr.9: 132727468-132727468 on Build GRCh38

View Full Product Details