Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98648305-98648566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
UniGene
Hs.552567
Gene Chromosome Location
Chr.12: 98645141-98735433 on Build GRCh38
Transcripts
NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1 XM_017019250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4662307 c.13G>C p.A5P Chr.12: 98648372-98648372 on Build GRCh38
5731582 c.58A>T p.I20F Chr.12: 98648417-98648417 on Build GRCh38
1639276 c.59T>C p.I20T Chr.12: 98648418-98648418 on Build GRCh38
6601836 c.105A>G p.L35L Chr.12: 98648464-98648464 on Build GRCh38
3368997 c.92G>A p.S31N Chr.12: 98648451-98648451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953501688 Chr.12: 98648339-98648339 on Build GRCh38
rs373035647 Chr.12: 98648397-98648397 on Build GRCh38
rs372259373 Chr.12: 98648468-98648468 on Build GRCh38
rs745436384 Chr.12: 98648376-98648376 on Build GRCh38
rs895445197 Chr.12: 98648529-98648529 on Build GRCh38
rs868444063 Chr.12: 98648420-98648420 on Build GRCh38
rs889955884 Chr.12: 98648398-98648398 on Build GRCh38
rs776606994 Chr.12: 98648375-98648375 on Build GRCh38
rs773776704 Chr.12: 98648523-98648523 on Build GRCh38
rs111232069 Chr.12: 98648349-98648349 on Build GRCh38
rs372096318 Chr.12: 98648520-98648520 on Build GRCh38
rs368174559 Chr.12: 98648417-98648417 on Build GRCh38
rs771167842 Chr.12: 98648456-98648456 on Build GRCh38
rs776731061 Chr.12: 98648475-98648475 on Build GRCh38
rs199944569 Chr.12: 98648495-98648495 on Build GRCh38
rs764496783 Chr.12: 98648535-98648535 on Build GRCh38
rs762601497 Chr.12: 98648407-98648407 on Build GRCh38
rs145226502 Chr.12: 98648493-98648493 on Build GRCh38
rs766035695 Chr.12: 98648521-98648521 on Build GRCh38
rs756605547 Chr.12: 98648534-98648534 on Build GRCh38
rs766952884 Chr.12: 98648527-98648527 on Build GRCh38
rs966090697 Chr.12: 98648396-98648396 on Build GRCh38
rs1016681790 Chr.12: 98648427-98648427 on Build GRCh38
rs759119224 Chr.12: 98648526-98648526 on Build GRCh38
rs1019334573 Chr.12: 98648394-98648394 on Build GRCh38
rs765077795 Chr.12: 98648382-98648382 on Build GRCh38
rs77355388 Chr.12: 98648519-98648519 on Build GRCh38
rs781643760 Chr.12: 98648431-98648431 on Build GRCh38
rs768615855 Chr.12: 98648359-98648359 on Build GRCh38
rs753165073 Chr.12: 98648435-98648435 on Build GRCh38
rs749575227 Chr.12: 98648452-98648452 on Build GRCh38
rs994258301 Chr.12: 98648455-98648455 on Build GRCh38
rs761730145 Chr.12: 98648381-98648381 on Build GRCh38
rs745929548 Chr.12: 98648467-98648467 on Build GRCh38
rs774512133 Chr.12: 98648336-98648337 on Build GRCh38
rs949748544 Chr.12: 98648532-98648532 on Build GRCh38
rs150457288 Chr.12: 98648430-98648430 on Build GRCh38
rs112605025 Chr.12: 98648505-98648505 on Build GRCh38
rs751071524 Chr.12: 98648426-98648426 on Build GRCh38
rs753203820 Chr.12: 98648530-98648530 on Build GRCh38
rs756482718 Chr.12: 98648451-98648451 on Build GRCh38
rs375102770 Chr.12: 98648500-98648500 on Build GRCh38
rs774480986 Chr.12: 98648350-98648350 on Build GRCh38
rs778908657 Chr.12: 98648457-98648457 on Build GRCh38
rs760587585 Chr.12: 98648353-98648353 on Build GRCh38

View Full Product Details