Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144520939-144521192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC14
Entrez Gene ID
9684
Gene Name
leucine rich repeat containing 14
Gene Aliases
LRRC14A
UniGene
Hs.459391
Gene Chromosome Location
Chr.8: 144517965-144525178 on Build GRCh38
Transcripts
NM_001272036.1 NM_014665.3 XM_005272358.4 XM_005272359.4 XM_005272360.4 XM_017014005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3898318 c.994A>G p.S332G Chr.8: 144520990-144520990 on Build GRCh38
3898319 c.1007C>A p.A336D Chr.8: 144521003-144521003 on Build GRCh38
5727784 c.1064C>T p.P355L Chr.8: 144521060-144521060 on Build GRCh38
7322781 c.981C>T p.R327R Chr.8: 144520977-144520977 on Build GRCh38
6536700 c.1027C>G p.L343V Chr.8: 144521023-144521023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962717015 Chr.8: 144521118-144521118 on Build GRCh38
rs761117107 Chr.8: 144520968-144520968 on Build GRCh38
rs750211969 Chr.8: 144521145-144521145 on Build GRCh38
rs756050011 Chr.8: 144521022-144521022 on Build GRCh38
rs775924504 Chr.8: 144521087-144521087 on Build GRCh38
rs200749025 Chr.8: 144521168-144521168 on Build GRCh38
rs773627094 Chr.8: 144520962-144520962 on Build GRCh38
rs115419329 Chr.8: 144521172-144521172 on Build GRCh38
rs763130431 Chr.8: 144521067-144521067 on Build GRCh38
rs760047081 Chr.8: 144520980-144520980 on Build GRCh38
rs754965671 Chr.8: 144521161-144521161 on Build GRCh38
rs757900139 Chr.8: 144520995-144520995 on Build GRCh38
rs1018713685 Chr.8: 144521097-144521097 on Build GRCh38
rs751142248 Chr.8: 144521007-144521007 on Build GRCh38
rs759864422 Chr.8: 144521081-144521081 on Build GRCh38
rs760735477 Chr.8: 144521115-144521118 on Build GRCh38
rs755921776 Chr.8: 144521151-144521151 on Build GRCh38
rs777103683 Chr.8: 144521073-144521073 on Build GRCh38
rs772316911 Chr.8: 144520960-144520960 on Build GRCh38
rs778012484 Chr.8: 144521061-144521061 on Build GRCh38
rs981308165 Chr.8: 144521081-144521083 on Build GRCh38
rs762276370 Chr.8: 144520987-144520987 on Build GRCh38
rs767200127 Chr.8: 144521140-144521140 on Build GRCh38
rs143508921 Chr.8: 144521034-144521034 on Build GRCh38
rs748247540 Chr.8: 144521035-144521035 on Build GRCh38
rs780920329 Chr.8: 144521016-144521016 on Build GRCh38
rs756920035 Chr.8: 144521013-144521013 on Build GRCh38
rs770317927 Chr.8: 144521086-144521086 on Build GRCh38
rs139683234 Chr.8: 144521000-144521000 on Build GRCh38
rs572054303 Chr.8: 144520975-144520975 on Build GRCh38
rs761417835 Chr.8: 144521136-144521136 on Build GRCh38
rs763542125 Chr.8: 144521110-144521110 on Build GRCh38
rs772111598 Chr.8: 144521041-144521041 on Build GRCh38
rs543083209 Chr.8: 144521160-144521160 on Build GRCh38
rs763516828 Chr.8: 144521106-144521106 on Build GRCh38
rs1007128616 Chr.8: 144521048-144521048 on Build GRCh38
rs780037818 Chr.8: 144521152-144521152 on Build GRCh38
rs145396273 Chr.8: 144521127-144521127 on Build GRCh38
rs773378779 Chr.8: 144521091-144521092 on Build GRCh38
rs752226395 Chr.8: 144520988-144520988 on Build GRCh38
rs750252014 Chr.8: 144521017-144521017 on Build GRCh38
rs150048017 Chr.8: 144521099-144521099 on Build GRCh38
rs370913027 Chr.8: 144521139-144521139 on Build GRCh38
rs747178364 Chr.8: 144521065-144521065 on Build GRCh38
rs765701739 Chr.8: 144520983-144520983 on Build GRCh38

View Full Product Details