Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55107994-55108255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMA
Entrez Gene ID
3001
Gene Name
granzyme A
Gene Aliases
CTLA3, HFSP
UniGene
Hs.90708
Gene Chromosome Location
Chr.5: 55102646-55110252 on Build GRCh38
Transcripts
NM_006144.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3381332 c.433G>T p.G145* Chr.5: 55108200-55108200 on Build GRCh38
7252735 c.462C>G p.G154G Chr.5: 55108229-55108229 on Build GRCh38
1253935 c.427delA p.K143fs*24 Chr.5: 55108194-55108194 on Build GRCh38
7216506 c.458G>T p.W153L Chr.5: 55108225-55108225 on Build GRCh38
4141997 c.372A>C p.A124A Chr.5: 55108139-55108139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559498444 Chr.5: 55108082-55108082 on Build GRCh38
rs753992275 Chr.5: 55108226-55108226 on Build GRCh38
rs369222212 Chr.5: 55108162-55108162 on Build GRCh38
rs977607957 Chr.5: 55108018-55108018 on Build GRCh38
rs372514056 Chr.5: 55108142-55108142 on Build GRCh38
rs762765865 Chr.5: 55108158-55108158 on Build GRCh38
rs552295796 Chr.5: 55108130-55108130 on Build GRCh38
rs904744139 Chr.5: 55108043-55108043 on Build GRCh38
rs1051846 Chr.5: 55108229-55108229 on Build GRCh38
rs530263188 Chr.5: 55108094-55108094 on Build GRCh38
rs779513892 Chr.5: 55108231-55108231 on Build GRCh38
rs1030106381 Chr.5: 55108228-55108228 on Build GRCh38
rs2270627 Chr.5: 55108225-55108225 on Build GRCh38
rs189633369 Chr.5: 55108100-55108100 on Build GRCh38
rs556099504 Chr.5: 55108190-55108190 on Build GRCh38
rs748574049 Chr.5: 55108108-55108108 on Build GRCh38
rs756084513 Chr.5: 55108079-55108079 on Build GRCh38
rs1875506 Chr.5: 55108187-55108187 on Build GRCh38
rs751516645 Chr.5: 55108168-55108168 on Build GRCh38
rs756420443 Chr.5: 55108201-55108201 on Build GRCh38
rs775130512 Chr.5: 55108156-55108156 on Build GRCh38
rs750590418 Chr.5: 55108200-55108200 on Build GRCh38
rs3104233 Chr.5: 55108129-55108129 on Build GRCh38
rs764138358 Chr.5: 55108164-55108164 on Build GRCh38
rs774550337 Chr.5: 55108106-55108106 on Build GRCh38
rs563675975 Chr.5: 55108119-55108119 on Build GRCh38
rs3104233 Chr.5: 55108129-55108129 on Build GRCh38

View Full Product Details