Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 64690641-64690874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM159B
Entrez Gene ID
100132916
Gene Name
family with sequence similarity 159 member B
Gene Aliases
-
UniGene
Hs.591776
Gene Chromosome Location
Chr.5: 64690308-64718190 on Build GRCh38
Transcripts
NM_001164442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408233 c.76C>T p.R26W Chr.5: 64690699-64690699 on Build GRCh38
1661752 c.94G>C p.A32P Chr.5: 64690717-64690717 on Build GRCh38
1661753 c.95C>T p.A32V Chr.5: 64690718-64690718 on Build GRCh38
6437451 c.117C>T p.F39F Chr.5: 64690740-64690740 on Build GRCh38
1735126 c.20T>C p.L7P Chr.5: 64690643-64690643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758318353 Chr.5: 64690757-64690757 on Build GRCh38
rs771873498 Chr.5: 64690801-64690801 on Build GRCh38
rs771008557 Chr.5: 64690765-64690765 on Build GRCh38
rs149547808 Chr.5: 64690689-64690689 on Build GRCh38
rs1019659113 Chr.5: 64690669-64690669 on Build GRCh38
rs967402995 Chr.5: 64690853-64690853 on Build GRCh38
rs776000409 Chr.5: 64690786-64690786 on Build GRCh38
rs746371062 Chr.5: 64690773-64690773 on Build GRCh38
rs756959762 Chr.5: 64690697-64690697 on Build GRCh38
rs576944799 Chr.5: 64690693-64690693 on Build GRCh38
rs1041778999 Chr.5: 64690711-64690711 on Build GRCh38
rs754789826 Chr.5: 64690740-64690740 on Build GRCh38
rs928365488 Chr.5: 64690713-64690713 on Build GRCh38
rs750213643 Chr.5: 64690735-64690735 on Build GRCh38
rs760152424 Chr.5: 64690804-64690804 on Build GRCh38
rs887027522 Chr.5: 64690764-64690764 on Build GRCh38
rs761550753 Chr.5: 64690699-64690699 on Build GRCh38
rs771193726 Chr.5: 64690668-64690668 on Build GRCh38
rs753416275 Chr.5: 64690695-64690695 on Build GRCh38
rs1045644522 Chr.5: 64690671-64690671 on Build GRCh38
rs961958768 Chr.5: 64690848-64690848 on Build GRCh38
rs528215913 Chr.5: 64690855-64690855 on Build GRCh38
rs1028380774 Chr.5: 64690833-64690833 on Build GRCh38
rs1028039935 Chr.5: 64690731-64690731 on Build GRCh38
rs1011331385 Chr.5: 64690730-64690730 on Build GRCh38
rs780716615 Chr.5: 64690695-64690695 on Build GRCh38
rs753787169 Chr.5: 64690821-64690821 on Build GRCh38
rs763352166 Chr.5: 64690791-64690791 on Build GRCh38
rs769606323 Chr.5: 64690729-64690729 on Build GRCh38
rs747127324 Chr.5: 64690761-64690761 on Build GRCh38
rs559454540 Chr.5: 64690810-64690810 on Build GRCh38
rs778766085 Chr.5: 64690749-64690749 on Build GRCh38
rs958169580 Chr.5: 64690834-64690834 on Build GRCh38
rs766268089 Chr.5: 64690708-64690708 on Build GRCh38
rs894290509 Chr.5: 64690734-64690734 on Build GRCh38
rs958263000 Chr.5: 64690694-64690694 on Build GRCh38
rs79949481 Chr.5: 64690707-64690707 on Build GRCh38
rs973007761 Chr.5: 64690857-64690857 on Build GRCh38
rs144658157 Chr.5: 64690782-64690782 on Build GRCh38
rs781594917 Chr.5: 64690767-64690767 on Build GRCh38
rs746881061 Chr.5: 64690796-64690796 on Build GRCh38
rs752425222 Chr.5: 64690751-64690751 on Build GRCh38
rs765901659 Chr.5: 64690819-64690819 on Build GRCh38
rs990060953 Chr.5: 64690841-64690841 on Build GRCh38
rs772968495 Chr.5: 64690802-64690802 on Build GRCh38
rs954206240 Chr.5: 64690769-64690769 on Build GRCh38
rs995761203 Chr.5: 64690747-64690747 on Build GRCh38
rs867977442 Chr.5: 64690742-64690742 on Build GRCh38
rs747037670 Chr.5: 64690678-64690678 on Build GRCh38
rs759524637 Chr.5: 64690838-64690838 on Build GRCh38

View Full Product Details