Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29828166-29828399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-G
Entrez Gene ID
3135
Gene Name
major histocompatibility complex, class I, G
Gene Aliases
MHC-G
UniGene
Hs.512152
Gene Chromosome Location
Chr.6: 29826967-29831130 on Build GRCh38
Transcripts
NM_002127.5 XM_017010816.1 XM_017010817.1 XM_017010818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
134116 c.203G>A p.R68K Chr.6: 29828176-29828176 on Build GRCh38
3861335 c.244G>A p.E82K Chr.6: 29828217-29828217 on Build GRCh38
4543515 c.339G>A p.E113E Chr.6: 29828312-29828312 on Build GRCh38
6351387 c.343+10T>C p.? Chr.6: 29828326-29828326 on Build GRCh38
6892053 c.253G>A p.E85K Chr.6: 29828226-29828226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776393668 Chr.6: 29828311-29828311 on Build GRCh38
rs773460671 Chr.6: 29828356-29828356 on Build GRCh38
rs41555713 Chr.6: 29828252-29828252 on Build GRCh38
rs41545515 Chr.6: 29828206-29828206 on Build GRCh38
rs757883678 Chr.6: 29828250-29828250 on Build GRCh38
rs766121990 Chr.6: 29828229-29828229 on Build GRCh38
rs200094914 Chr.6: 29828212-29828212 on Build GRCh38
rs756969252 Chr.6: 29828261-29828261 on Build GRCh38
rs766321859 Chr.6: 29828354-29828358 on Build GRCh38
rs752196294 Chr.6: 29828248-29828248 on Build GRCh38
rs201510147 Chr.6: 29828342-29828342 on Build GRCh38
rs1736927 Chr.6: 29828338-29828338 on Build GRCh38
rs113688647 Chr.6: 29828318-29828318 on Build GRCh38
rs746249960 Chr.6: 29828266-29828266 on Build GRCh38
rs17875399 Chr.6: 29828329-29828329 on Build GRCh38
rs756802655 Chr.6: 29828333-29828333 on Build GRCh38
rs751127247 Chr.6: 29828253-29828253 on Build GRCh38
rs557373005 Chr.6: 29828372-29828372 on Build GRCh38
rs781049802 Chr.6: 29828334-29828334 on Build GRCh38
rs759390427 Chr.6: 29828312-29828312 on Build GRCh38
rs778800327 Chr.6: 29828345-29828345 on Build GRCh38
rs79178226 Chr.6: 29828205-29828205 on Build GRCh38
rs774484424 Chr.6: 29828222-29828222 on Build GRCh38
rs768569399 Chr.6: 29828300-29828300 on Build GRCh38
rs954164331 Chr.6: 29828267-29828267 on Build GRCh38
rs1626038 Chr.6: 29828326-29828326 on Build GRCh38
rs17875400 Chr.6: 29828351-29828351 on Build GRCh38
rs369333018 Chr.6: 29828354-29828355 on Build GRCh38
rs9280715 Chr.6: 29828350-29828351 on Build GRCh38
rs770412396 Chr.6: 29828278-29828278 on Build GRCh38
rs373168953 Chr.6: 29828292-29828292 on Build GRCh38
rs770430489 Chr.6: 29828192-29828192 on Build GRCh38
rs772273052 Chr.6: 29828355-29828355 on Build GRCh38
rs761874800 Chr.6: 29828310-29828310 on Build GRCh38
rs1052344302 Chr.6: 29828379-29828379 on Build GRCh38
rs1130355 Chr.6: 29828216-29828216 on Build GRCh38
rs765213288 Chr.6: 29828239-29828239 on Build GRCh38
rs373197901 Chr.6: 29828226-29828226 on Build GRCh38
rs752628469 Chr.6: 29828325-29828325 on Build GRCh38
rs111241895 Chr.6: 29828314-29828314 on Build GRCh38
rs535263603 Chr.6: 29828337-29828337 on Build GRCh38
rs1625907 Chr.6: 29828375-29828375 on Build GRCh38
rs1046196885 Chr.6: 29828357-29828357 on Build GRCh38
rs775653233 Chr.6: 29828193-29828193 on Build GRCh38
rs781636203 Chr.6: 29828263-29828263 on Build GRCh38
rs78627024 Chr.6: 29828210-29828210 on Build GRCh38
rs780775783 Chr.6: 29828289-29828289 on Build GRCh38
rs3215482 Chr.6: 29828349-29828350 on Build GRCh38
rs753634137 Chr.6: 29828238-29828238 on Build GRCh38
rs780585877 Chr.6: 29828341-29828341 on Build GRCh38
rs113311833 Chr.6: 29828228-29828228 on Build GRCh38

View Full Product Details