Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74632771-74632996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK3
Entrez Gene ID
1198
Gene Name
CDC like kinase 3
Gene Aliases
PHCLK3, PHCLK3/152
UniGene
Hs.511790
Gene Chromosome Location
Chr.15: 74598476-74645418 on Build GRCh38
Transcripts
NM_001142443.1 NM_001142444.1 NM_025083.3 XM_011522092.2

Gene Symbol
EDC3
Entrez Gene ID
80153
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
LSM16, MRT50, YJDC, YJEFN2
UniGene
Hs.682454
Gene Chromosome Location
Chr.15: 74630558-74696045 on Build GRCh38
Transcripts
NM_001142443.1 NM_001142444.1 NM_025083.3 XM_011522092.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1250924 c.1217delT p.L406fs*37 Chr.15: 74632922-74632922 on Build GRCh38
5513112 c.1306C>T p.R436W Chr.15: 74632833-74632833 on Build GRCh38
6341106 c.1313C>A p.P438Q Chr.15: 74632826-74632826 on Build GRCh38
7326911 c.1329_1330ins57 p.D443_P444ins19 Chr.15: 74632809-74632810 on Build GRCh38
5549863 c.1310C>T p.A437V Chr.15: 74632829-74632829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750199559 Chr.15: 74632977-74632977 on Build GRCh38
rs780034536 Chr.15: 74632810-74632810 on Build GRCh38
rs548586544 Chr.15: 74632792-74632792 on Build GRCh38
rs776955975 Chr.15: 74632891-74632891 on Build GRCh38
rs773653347 Chr.15: 74632975-74632976 on Build GRCh38
rs112697815 Chr.15: 74632799-74632799 on Build GRCh38
rs756054202 Chr.15: 74632801-74632801 on Build GRCh38
rs1012516724 Chr.15: 74632881-74632881 on Build GRCh38
rs368247768 Chr.15: 74632892-74632892 on Build GRCh38
rs1053944795 Chr.15: 74632825-74632825 on Build GRCh38
rs148691547 Chr.15: 74632793-74632793 on Build GRCh38
rs761542454 Chr.15: 74632927-74632927 on Build GRCh38
rs774213135 Chr.15: 74632908-74632908 on Build GRCh38
rs747512793 Chr.15: 74632880-74632880 on Build GRCh38
rs773607334 Chr.15: 74632879-74632879 on Build GRCh38
rs893947456 Chr.15: 74632828-74632828 on Build GRCh38
rs371061282 Chr.15: 74632815-74632815 on Build GRCh38
rs754998831 Chr.15: 74632843-74632843 on Build GRCh38
rs772486484 Chr.15: 74632878-74632878 on Build GRCh38
rs556849496 Chr.15: 74632972-74632972 on Build GRCh38
rs999717700 Chr.15: 74632824-74632824 on Build GRCh38
rs547318062 Chr.15: 74632809-74632809 on Build GRCh38
rs767569596 Chr.15: 74632912-74632912 on Build GRCh38
rs372344016 Chr.15: 74632900-74632900 on Build GRCh38
rs779787138 Chr.15: 74632909-74632909 on Build GRCh38
rs367953733 Chr.15: 74632791-74632791 on Build GRCh38
rs767468142 Chr.15: 74632967-74632967 on Build GRCh38
rs748368878 Chr.15: 74632867-74632867 on Build GRCh38
rs375577225 Chr.15: 74632890-74632890 on Build GRCh38

View Full Product Details