Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181232492-181232763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM41
Entrez Gene ID
90933
Gene Name
tripartite motif containing 41
Gene Aliases
RINCK
UniGene
Hs.441488
Gene Chromosome Location
Chr.5: 181223263-181235808 on Build GRCh38
Transcripts
NM_033549.4 NM_201627.2 NR_045218.1 XM_006714929.2 XM_006714930.2 XM_006714931.3 XM_006714932.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3615052 c.956G>A p.R319Q Chr.5: 181232705-181232705 on Build GRCh38
7138539 c.960G>C p.L320L Chr.5: 181232709-181232709 on Build GRCh38
3696847 c.918G>T p.M306I Chr.5: 181232667-181232667 on Build GRCh38
3696846 c.918G>T p.M306I Chr.5: 181232667-181232667 on Build GRCh38
7138538 c.960G>C p.L320L Chr.5: 181232709-181232709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772071153 Chr.5: 181232614-181232614 on Build GRCh38
rs969278833 Chr.5: 181232589-181232589 on Build GRCh38
rs772243270 Chr.5: 181232644-181232646 on Build GRCh38
rs944870478 Chr.5: 181232549-181232549 on Build GRCh38
rs761638995 Chr.5: 181232661-181232661 on Build GRCh38
rs753810828 Chr.5: 181232693-181232693 on Build GRCh38
rs746936559 Chr.5: 181232618-181232618 on Build GRCh38
rs756574391 Chr.5: 181232714-181232714 on Build GRCh38
rs763850506 Chr.5: 181232531-181232531 on Build GRCh38
rs992230319 Chr.5: 181232579-181232579 on Build GRCh38
rs1045661463 Chr.5: 181232656-181232656 on Build GRCh38
rs117893497 Chr.5: 181232540-181232540 on Build GRCh38
rs369269974 Chr.5: 181232705-181232705 on Build GRCh38
rs948350857 Chr.5: 181232648-181232648 on Build GRCh38
rs181154343 Chr.5: 181232617-181232617 on Build GRCh38
rs1015096883 Chr.5: 181232546-181232546 on Build GRCh38
rs772763077 Chr.5: 181232685-181232685 on Build GRCh38
rs745909348 Chr.5: 181232613-181232613 on Build GRCh38
rs764736502 Chr.5: 181232736-181232736 on Build GRCh38
rs189635310 Chr.5: 181232543-181232543 on Build GRCh38
rs768642232 Chr.5: 181232655-181232655 on Build GRCh38
rs568397611 Chr.5: 181232567-181232567 on Build GRCh38
rs386695879 Chr.5: 181232530-181232532 on Build GRCh38
rs913965930 Chr.5: 181232730-181232730 on Build GRCh38
rs777703507 Chr.5: 181232648-181232648 on Build GRCh38
rs977615727 Chr.5: 181232571-181232571 on Build GRCh38
rs80188051 Chr.5: 181232530-181232530 on Build GRCh38
rs924754923 Chr.5: 181232586-181232586 on Build GRCh38
rs113274889 Chr.5: 181232604-181232604 on Build GRCh38
rs139611453 Chr.5: 181232659-181232659 on Build GRCh38
rs1015365925 Chr.5: 181232528-181232528 on Build GRCh38
rs1015421315 Chr.5: 181232725-181232725 on Build GRCh38
rs534188943 Chr.5: 181232588-181232588 on Build GRCh38
rs551287324 Chr.5: 181232609-181232609 on Build GRCh38
rs150479742 Chr.5: 181232532-181232532 on Build GRCh38
rs532360737 Chr.5: 181232517-181232517 on Build GRCh38
rs752175226 Chr.5: 181232538-181232538 on Build GRCh38
rs757215614 Chr.5: 181232631-181232631 on Build GRCh38
rs765812251 Chr.5: 181232707-181232707 on Build GRCh38
rs751138682 Chr.5: 181232711-181232711 on Build GRCh38
rs764848597 Chr.5: 181232684-181232684 on Build GRCh38
rs373867218 Chr.5: 181232610-181232610 on Build GRCh38
rs936503470 Chr.5: 181232644-181232644 on Build GRCh38

View Full Product Details