Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178390019-178390286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASAL2
Entrez Gene ID
9462
Gene Name
RAS protein activator like 2
Gene Aliases
NGAP
UniGene
Hs.496139
Gene Chromosome Location
Chr.1: 178093729-178479514 on Build GRCh38
Transcripts
NM_004841.3 NM_170692.2 XM_005245622.3 XM_011510166.2 XM_011510167.2 XM_017002849.1 XM_017002850.1 XM_017002851.1 XM_017002852.1 XM_017002853.1 XM_017002854.1 XM_017002855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5745757 c.493T>C p.S165P Chr.1: 178390189-178390189 on Build GRCh38
5745758 c.103T>C p.S35P Chr.1: 178390189-178390189 on Build GRCh38
5442625 c.104C>T p.S35L Chr.1: 178390190-178390190 on Build GRCh38
125766 c.473C>T p.S158F Chr.1: 178390169-178390169 on Build GRCh38
5442622 c.53C>T p.S18L Chr.1: 178390139-178390139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763722066 Chr.1: 178390229-178390229 on Build GRCh38
rs780202107 Chr.1: 178390164-178390164 on Build GRCh38
rs759227860 Chr.1: 178390123-178390123 on Build GRCh38
rs923084923 Chr.1: 178390196-178390196 on Build GRCh38
rs755054848 Chr.1: 178390171-178390171 on Build GRCh38
rs977977263 Chr.1: 178390240-178390240 on Build GRCh38
rs752286353 Chr.1: 178390092-178390092 on Build GRCh38
rs767521333 Chr.1: 178390247-178390247 on Build GRCh38
rs201452413 Chr.1: 178390130-178390130 on Build GRCh38
rs769464413 Chr.1: 178390204-178390204 on Build GRCh38
rs558228744 Chr.1: 178390080-178390080 on Build GRCh38
rs143298701 Chr.1: 178390048-178390048 on Build GRCh38
rs936599431 Chr.1: 178390260-178390260 on Build GRCh38
rs748664966 Chr.1: 178390173-178390173 on Build GRCh38
rs764742724 Chr.1: 178390206-178390206 on Build GRCh38
rs371914361 Chr.1: 178390115-178390115 on Build GRCh38
rs773557078 Chr.1: 178390186-178390186 on Build GRCh38
rs774374898 Chr.1: 178390230-178390230 on Build GRCh38
rs965136061 Chr.1: 178390167-178390167 on Build GRCh38
rs763501844 Chr.1: 178390178-178390178 on Build GRCh38
rs772352575 Chr.1: 178390177-178390177 on Build GRCh38
rs762483066 Chr.1: 178390226-178390226 on Build GRCh38
rs374544024 Chr.1: 178390245-178390245 on Build GRCh38
rs762418560 Chr.1: 178390129-178390129 on Build GRCh38
rs764773286 Chr.1: 178390047-178390047 on Build GRCh38
rs375225072 Chr.1: 178390140-178390140 on Build GRCh38
rs776204638 Chr.1: 178390116-178390116 on Build GRCh38
rs933192811 Chr.1: 178390205-178390205 on Build GRCh38
rs747390240 Chr.1: 178390192-178390192 on Build GRCh38
rs779341777 Chr.1: 178390172-178390172 on Build GRCh38
rs750224018 Chr.1: 178390258-178390258 on Build GRCh38
rs774890628 Chr.1: 178390109-178390109 on Build GRCh38
rs950690151 Chr.1: 178390101-178390101 on Build GRCh38
rs775050848 Chr.1: 178390127-178390127 on Build GRCh38
rs762123709 Chr.1: 178390111-178390111 on Build GRCh38
rs769196546 Chr.1: 178390105-178390105 on Build GRCh38
rs200536697 Chr.1: 178390169-178390169 on Build GRCh38
rs757071448 Chr.1: 178390150-178390150 on Build GRCh38
rs777628208 Chr.1: 178390049-178390049 on Build GRCh38
rs761847039 Chr.1: 178390241-178390241 on Build GRCh38
rs746116717 Chr.1: 178390070-178390070 on Build GRCh38
rs764865700 Chr.1: 178390124-178390124 on Build GRCh38
rs903944749 Chr.1: 178390063-178390075 on Build GRCh38
rs1020990870 Chr.1: 178390093-178390093 on Build GRCh38
rs958754238 Chr.1: 178390225-178390225 on Build GRCh38
rs767433897 Chr.1: 178390151-178390151 on Build GRCh38
rs781526075 Chr.1: 178390058-178390058 on Build GRCh38
rs756359216 Chr.1: 178390156-178390156 on Build GRCh38
rs749398586 Chr.1: 178390082-178390082 on Build GRCh38
rs757155612 Chr.1: 178390056-178390056 on Build GRCh38
rs751376776 Chr.1: 178390054-178390054 on Build GRCh38

View Full Product Details