Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145849703-145849442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIAS3
Entrez Gene ID
10401
Gene Name
protein inhibitor of activated STAT 3
Gene Aliases
ZMIZ5
UniGene
Hs.435761
Gene Chromosome Location
Chr.1: 145848522-145859081 on Build GRCh38
Transcripts
NM_006099.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4473353 c.1844C>T p.P615L Chr.1: 145849489-145849489 on Build GRCh38
5923542 c.1739C>T p.S580F Chr.1: 145849594-145849594 on Build GRCh38
1180694 c.1798delG p.A602fs*2 Chr.1: 145849535-145849535 on Build GRCh38
4473337 c.1843C>T p.P615S Chr.1: 145849490-145849490 on Build GRCh38
1220520 c.1759G>A p.A587T Chr.1: 145849574-145849574 on Build GRCh38
View More

View Full Product Details