Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51945839-51946102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP3
Entrez Gene ID
10039
Gene Name
poly(ADP-ribose) polymerase family member 3
Gene Aliases
ADPRT3, ADPRTL2, ADPRTL3, ARTD3, IRT1, PADPRT-3
UniGene
Hs.271742
Gene Chromosome Location
Chr.3: 51942282-51948867 on Build GRCh38
Transcripts
NM_001003931.3 NM_005485.5 XM_005264779.4 XM_017005490.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1663322 c.1050C>A p.H350Q Chr.3: 51945891-51945891 on Build GRCh38
7310707 c.1058C>A p.P353H Chr.3: 51945899-51945899 on Build GRCh38
7310706 c.1079C>A p.P360H Chr.3: 51945899-51945899 on Build GRCh38
6338802 c.1012-5C>T p.? Chr.3: 51945848-51945848 on Build GRCh38
1579451 c.1117G>A p.E373K Chr.3: 51945937-51945937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945003577 Chr.3: 51946039-51946039 on Build GRCh38
rs754741701 Chr.3: 51945923-51945923 on Build GRCh38
rs550999771 Chr.3: 51945915-51945915 on Build GRCh38
rs770922198 Chr.3: 51945953-51945953 on Build GRCh38
rs765281740 Chr.3: 51945965-51945965 on Build GRCh38
rs753433682 Chr.3: 51945919-51945919 on Build GRCh38
rs908379040 Chr.3: 51945942-51945942 on Build GRCh38
rs908141457 Chr.3: 51945984-51945984 on Build GRCh38
rs748058853 Chr.3: 51945929-51945929 on Build GRCh38
rs759879310 Chr.3: 51945896-51945896 on Build GRCh38
rs199581414 Chr.3: 51945859-51945859 on Build GRCh38
rs764574573 Chr.3: 51945989-51945989 on Build GRCh38
rs752575345 Chr.3: 51946048-51946048 on Build GRCh38
rs371686193 Chr.3: 51945967-51945967 on Build GRCh38
rs772102477 Chr.3: 51945938-51945938 on Build GRCh38
rs199987180 Chr.3: 51945955-51945955 on Build GRCh38
rs747082212 Chr.3: 51945952-51945952 on Build GRCh38
rs904087535 Chr.3: 51946035-51946035 on Build GRCh38
rs770158404 Chr.3: 51945959-51945959 on Build GRCh38
rs200569026 Chr.3: 51945903-51945903 on Build GRCh38
rs867059522 Chr.3: 51946061-51946061 on Build GRCh38
rs373700583 Chr.3: 51946006-51946006 on Build GRCh38
rs114378734 Chr.3: 51946063-51946063 on Build GRCh38
rs759700559 Chr.3: 51945956-51945956 on Build GRCh38
rs375151260 Chr.3: 51945982-51945982 on Build GRCh38
rs753203252 Chr.3: 51945899-51945899 on Build GRCh38
rs948416306 Chr.3: 51946005-51946005 on Build GRCh38
rs778594031 Chr.3: 51945927-51945927 on Build GRCh38
rs200205049 Chr.3: 51945946-51945946 on Build GRCh38
rs761707566 Chr.3: 51945886-51945886 on Build GRCh38
rs755676813 Chr.3: 51945918-51945918 on Build GRCh38
rs764627892 Chr.3: 51945912-51945912 on Build GRCh38
rs759691384 Chr.3: 51946049-51946049 on Build GRCh38
rs41292348 Chr.3: 51946060-51946060 on Build GRCh38
rs983693602 Chr.3: 51945972-51945972 on Build GRCh38
rs765533970 Chr.3: 51945898-51945898 on Build GRCh38
rs759386901 Chr.3: 51945892-51945892 on Build GRCh38

View Full Product Details