Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56166173-56166434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAG1
Entrez Gene ID
5324
Gene Name
PLAG1 zinc finger
Gene Aliases
PSA, SGPA, ZNF912
UniGene
Hs.14968
Gene Chromosome Location
Chr.8: 56160909-56211327 on Build GRCh38
Transcripts
NM_001114634.1 NM_001114635.1 NM_002655.2 XM_011517544.2 XM_017013576.1 XM_017013577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100658 c.1362C>T p.S454S Chr.8: 56166384-56166384 on Build GRCh38
5874832 c.1493C>T p.A498V Chr.8: 56166253-56166253 on Build GRCh38
1100656 c.1480C>T p.R494C Chr.8: 56166266-56166266 on Build GRCh38
3925422 c.1396C>T p.P466S Chr.8: 56166350-56166350 on Build GRCh38
751126 c.1359T>A p.V453V Chr.8: 56166387-56166387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780957066 Chr.8: 56166327-56166332 on Build GRCh38
rs141535675 Chr.8: 56166359-56166359 on Build GRCh38
rs904239542 Chr.8: 56166213-56166213 on Build GRCh38
rs559524753 Chr.8: 56166338-56166338 on Build GRCh38
rs75108631 Chr.8: 56166389-56166389 on Build GRCh38
rs764507623 Chr.8: 56166266-56166266 on Build GRCh38
rs188076400 Chr.8: 56166387-56166387 on Build GRCh38
rs759676950 Chr.8: 56166241-56166241 on Build GRCh38
rs768369873 Chr.8: 56166202-56166202 on Build GRCh38
rs1014167624 Chr.8: 56166345-56166345 on Build GRCh38
rs754664950 Chr.8: 56166412-56166412 on Build GRCh38
rs765825114 Chr.8: 56166400-56166400 on Build GRCh38
rs750003428 Chr.8: 56166314-56166314 on Build GRCh38
rs761135682 Chr.8: 56166265-56166265 on Build GRCh38
rs764795534 Chr.8: 56166208-56166208 on Build GRCh38
rs548976190 Chr.8: 56166321-56166321 on Build GRCh38
rs144552372 Chr.8: 56166373-56166373 on Build GRCh38
rs780999802 Chr.8: 56166354-56166354 on Build GRCh38
rs182997597 Chr.8: 56166255-56166255 on Build GRCh38
rs770946747 Chr.8: 56166231-56166231 on Build GRCh38
rs765836089 Chr.8: 56166391-56166391 on Build GRCh38
rs750826262 Chr.8: 56166397-56166397 on Build GRCh38
rs769829087 Chr.8: 56166368-56166368 on Build GRCh38
rs151260595 Chr.8: 56166278-56166278 on Build GRCh38
rs779683273 Chr.8: 56166324-56166324 on Build GRCh38
rs747864315 Chr.8: 56166355-56166355 on Build GRCh38
rs534097674 Chr.8: 56166228-56166228 on Build GRCh38
rs774529289 Chr.8: 56166238-56166238 on Build GRCh38
rs751242480 Chr.8: 56166402-56166402 on Build GRCh38
rs529202300 Chr.8: 56166270-56166270 on Build GRCh38
rs764195832 Chr.8: 56166378-56166378 on Build GRCh38
rs746604920 Chr.8: 56166337-56166337 on Build GRCh38
rs754956552 Chr.8: 56166211-56166211 on Build GRCh38
rs757915002 Chr.8: 56166398-56166398 on Build GRCh38
rs35883156 Chr.8: 56166374-56166374 on Build GRCh38
rs779262710 Chr.8: 56166286-56166286 on Build GRCh38

View Full Product Details