Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48713096-48713369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMSTR
Entrez Gene ID
284358
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
MASTR
UniGene
Hs.191815
Gene Chromosome Location
Chr.19: 48705718-48719898 on Build GRCh38
Transcripts
NM_001130915.1 NM_001297753.1 NM_182574.2 XM_011526807.2 XM_011526808.2 XM_011526809.2 XM_017026640.1 XM_017026641.1 XM_017026642.1 XM_017026643.1 XM_017026644.1 XM_017026645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1257200 c.1244G>A p.W415* Chr.19: 48713271-48713271 on Build GRCh38
3107798 c.851C>A p.S284Y Chr.19: 48713355-48713355 on Build GRCh38
6706581 c.867delC p.S290fs*>23 Chr.19: 48713339-48713339 on Build GRCh38
4462329 c.932C>T p.P311L Chr.19: 48713274-48713274 on Build GRCh38
6706579 c.910C>T p.L304L Chr.19: 48713296-48713296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770909901 Chr.19: 48713314-48713314 on Build GRCh38
rs760913677 Chr.19: 48713347-48713347 on Build GRCh38
rs746752773 Chr.19: 48713291-48713291 on Build GRCh38
rs761265109 Chr.19: 48713230-48713230 on Build GRCh38
rs186220936 Chr.19: 48713215-48713215 on Build GRCh38
rs759958885 Chr.19: 48713223-48713223 on Build GRCh38
rs374547209 Chr.19: 48713281-48713281 on Build GRCh38
rs200133856 Chr.19: 48713286-48713286 on Build GRCh38
rs765001875 Chr.19: 48713268-48713268 on Build GRCh38
rs911750073 Chr.19: 48713214-48713214 on Build GRCh38
rs868521623 Chr.19: 48713340-48713340 on Build GRCh38
rs775772010 Chr.19: 48713228-48713228 on Build GRCh38
rs750098803 Chr.19: 48713280-48713280 on Build GRCh38
rs985940684 Chr.19: 48713162-48713162 on Build GRCh38
rs373915683 Chr.19: 48713294-48713294 on Build GRCh38
rs761990111 Chr.19: 48713225-48713225 on Build GRCh38
rs749442911 Chr.19: 48713299-48713299 on Build GRCh38
rs746101267 Chr.19: 48713321-48713321 on Build GRCh38
rs753424243 Chr.19: 48713250-48713250 on Build GRCh38
rs774347340 Chr.19: 48713317-48713317 on Build GRCh38
rs556967114 Chr.19: 48713326-48713326 on Build GRCh38
rs926630531 Chr.19: 48713342-48713342 on Build GRCh38
rs774542969 Chr.19: 48713221-48713221 on Build GRCh38
rs779689325 Chr.19: 48713245-48713246 on Build GRCh38
rs750489528 Chr.19: 48713153-48713153 on Build GRCh38
rs1004265289 Chr.19: 48713220-48713220 on Build GRCh38
rs780968657 Chr.19: 48713295-48713295 on Build GRCh38
rs73057542 Chr.19: 48713350-48713350 on Build GRCh38
rs1029811691 Chr.19: 48713164-48713164 on Build GRCh38
rs756702812 Chr.19: 48713256-48713256 on Build GRCh38
rs958626978 Chr.19: 48713346-48713346 on Build GRCh38
rs963144199 Chr.19: 48713298-48713298 on Build GRCh38
rs1015679462 Chr.19: 48713255-48713255 on Build GRCh38
rs550062222 Chr.19: 48713344-48713344 on Build GRCh38

View Full Product Details