Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36963005-36963264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI16
Entrez Gene ID
221476
Gene Name
peptidase inhibitor 16
Gene Aliases
CD364, CRISP9, MSMBBP, PSPBP
UniGene
Hs.25391
Gene Chromosome Location
Chr.6: 36948263-36964837 on Build GRCh38
Transcripts
NM_001199159.1 NM_153370.2 XM_005248917.2 XM_011514375.2 XM_017010430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7105536 c.810G>T p.T270T Chr.6: 36963152-36963152 on Build GRCh38
5720593 c.837G>A p.E279E Chr.6: 36963179-36963179 on Build GRCh38
7121738 c.672A>T p.A224A Chr.6: 36963014-36963014 on Build GRCh38
7121739 c.680C>G p.S227C Chr.6: 36963022-36963022 on Build GRCh38
451369 c.864C>A p.V288V Chr.6: 36963206-36963206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763232029 Chr.6: 36963161-36963161 on Build GRCh38
rs141551770 Chr.6: 36963216-36963216 on Build GRCh38
rs927816754 Chr.6: 36963114-36963114 on Build GRCh38
rs1040603398 Chr.6: 36963243-36963243 on Build GRCh38
rs555165762 Chr.6: 36963173-36963173 on Build GRCh38
rs770420658 Chr.6: 36963134-36963134 on Build GRCh38
rs769926977 Chr.6: 36963207-36963207 on Build GRCh38
rs763465920 Chr.6: 36963219-36963219 on Build GRCh38
rs113296030 Chr.6: 36963228-36963228 on Build GRCh38
rs773312881 Chr.6: 36963075-36963075 on Build GRCh38
rs761500620 Chr.6: 36963184-36963184 on Build GRCh38
rs146645609 Chr.6: 36963191-36963191 on Build GRCh38
rs764477574 Chr.6: 36963163-36963163 on Build GRCh38
rs757363610 Chr.6: 36963077-36963077 on Build GRCh38
rs1049773721 Chr.6: 36963120-36963120 on Build GRCh38
rs747217941 Chr.6: 36963105-36963105 on Build GRCh38
rs868599175 Chr.6: 36963220-36963220 on Build GRCh38
rs778723339 Chr.6: 36963198-36963198 on Build GRCh38
rs368712205 Chr.6: 36963230-36963230 on Build GRCh38
rs922575539 Chr.6: 36963059-36963059 on Build GRCh38
rs779887710 Chr.6: 36963056-36963056 on Build GRCh38
rs753712773 Chr.6: 36963194-36963194 on Build GRCh38
rs760187412 Chr.6: 36963238-36963238 on Build GRCh38
rs535616461 Chr.6: 36963121-36963121 on Build GRCh38
rs150370433 Chr.6: 36963111-36963111 on Build GRCh38
rs372057432 Chr.6: 36963240-36963240 on Build GRCh38
rs75061272 Chr.6: 36963205-36963205 on Build GRCh38
rs145022350 Chr.6: 36963126-36963126 on Build GRCh38
rs756012600 Chr.6: 36963045-36963045 on Build GRCh38
rs969255840 Chr.6: 36963094-36963094 on Build GRCh38
rs532101741 Chr.6: 36963233-36963233 on Build GRCh38
rs780703150 Chr.6: 36963037-36963037 on Build GRCh38
rs776082173 Chr.6: 36963155-36963155 on Build GRCh38
rs767139014 Chr.6: 36963236-36963236 on Build GRCh38
rs908718134 Chr.6: 36963034-36963034 on Build GRCh38
rs41272198 Chr.6: 36963038-36963038 on Build GRCh38
rs754763464 Chr.6: 36963196-36963196 on Build GRCh38
rs776831640 Chr.6: 36963116-36963116 on Build GRCh38
rs199875004 Chr.6: 36963192-36963192 on Build GRCh38
rs867204264 Chr.6: 36963074-36963074 on Build GRCh38
rs771155764 Chr.6: 36963201-36963201 on Build GRCh38
rs139089988 Chr.6: 36963200-36963200 on Build GRCh38
rs747267930 Chr.6: 36963040-36963042 on Build GRCh38
rs768949758 Chr.6: 36963226-36963226 on Build GRCh38
rs768444413 Chr.6: 36963061-36963061 on Build GRCh38

View Full Product Details