Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139341286-139341531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf55-LUC7L2
Entrez Gene ID
100996928
Gene Name
C7orf55-LUC7L2 readthrough
Gene Aliases
-
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139341132-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_197964.4 NR_073058.1 NR_073059.1 NR_073060.1 NM_001244585.1

Gene Symbol
FMC1
Entrez Gene ID
154791
Gene Name
formation of mitochondrial complex V assembly factor 1 homolog
Gene Aliases
C7orf55, HSPC268
UniGene
Hs.718441
Gene Chromosome Location
Chr.7: 139340450-139346319 on Build GRCh38
Transcripts
NM_001244584.2 NM_197964.4 NR_073058.1 NR_073059.1 NR_073060.1 NM_001244585.1

Gene Symbol
LUC7L2
Entrez Gene ID
51631
Gene Name
LUC7 like 2, pre-mRNA splicing factor
Gene Aliases
CGI-59, CGI-74, LUC7B2
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139340359-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_197964.4 NR_073058.1 NR_073059.1 NR_073060.1 NM_001244585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5580461 c.110A>G p.Y37C Chr.7: 139341494-139341494 on Build GRCh38
1312731 c.106C>G p.R36G Chr.7: 139341490-139341490 on Build GRCh38
4942282 c.9C>T p.A3A Chr.7: 139341393-139341393 on Build GRCh38
6515666 c.56G>A p.R19H Chr.7: 139341440-139341440 on Build GRCh38
4389824 c.72C>T p.A24A Chr.7: 139341456-139341456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781571120 Chr.7: 139341361-139341361 on Build GRCh38
rs756177561 Chr.7: 139341508-139341508 on Build GRCh38
rs571616041 Chr.7: 139341500-139341500 on Build GRCh38
rs762406780 Chr.7: 139341488-139341488 on Build GRCh38
rs778569166 Chr.7: 139341384-139341384 on Build GRCh38
rs775057665 Chr.7: 139341484-139341484 on Build GRCh38
rs928270619 Chr.7: 139341329-139341329 on Build GRCh38
rs758182186 Chr.7: 139341512-139341512 on Build GRCh38
rs546646905 Chr.7: 139341470-139341470 on Build GRCh38
rs746030576 Chr.7: 139341364-139341364 on Build GRCh38
rs759231210 Chr.7: 139341452-139341452 on Build GRCh38
rs375052584 Chr.7: 139341413-139341413 on Build GRCh38
rs763234210 Chr.7: 139341480-139341480 on Build GRCh38
rs199542596 Chr.7: 139341418-139341418 on Build GRCh38
rs779672664 Chr.7: 139341321-139341321 on Build GRCh38
rs201970386 Chr.7: 139341431-139341431 on Build GRCh38
rs747925343 Chr.7: 139341416-139341416 on Build GRCh38
rs139756425 Chr.7: 139341440-139341440 on Build GRCh38
rs146144422 Chr.7: 139341503-139341503 on Build GRCh38
rs771925654 Chr.7: 139341385-139341385 on Build GRCh38
rs756259682 Chr.7: 139341412-139341412 on Build GRCh38
rs769050200 Chr.7: 139341467-139341467 on Build GRCh38
rs751714776 Chr.7: 139341342-139341342 on Build GRCh38
rs141486673 Chr.7: 139341383-139341383 on Build GRCh38
rs544455569 Chr.7: 139341490-139341490 on Build GRCh38
rs10265 Chr.7: 139341406-139341406 on Build GRCh38
rs372818202 Chr.7: 139341353-139341353 on Build GRCh38
rs1055740079 Chr.7: 139341369-139341369 on Build GRCh38
rs369452273 Chr.7: 139341459-139341459 on Build GRCh38
rs559843930 Chr.7: 139341371-139341373 on Build GRCh38
rs746933781 Chr.7: 139341435-139341435 on Build GRCh38
rs114009688 Chr.7: 139341333-139341333 on Build GRCh38
rs752049299 Chr.7: 139341404-139341404 on Build GRCh38
rs754031356 Chr.7: 139341511-139341511 on Build GRCh38
rs776084824 Chr.7: 139341444-139341444 on Build GRCh38
rs773724554 Chr.7: 139341338-139341338 on Build GRCh38
rs563299778 Chr.7: 139341390-139341390 on Build GRCh38
rs777537487 Chr.7: 139341514-139341514 on Build GRCh38
rs980567869 Chr.7: 139341506-139341506 on Build GRCh38
rs530614678 Chr.7: 139341393-139341393 on Build GRCh38
rs775171261 Chr.7: 139341458-139341458 on Build GRCh38
rs769358474 Chr.7: 139341456-139341456 on Build GRCh38
rs764543305 Chr.7: 139341403-139341403 on Build GRCh38
rs771832282 Chr.7: 139341421-139341421 on Build GRCh38
rs757375443 Chr.7: 139341356-139341356 on Build GRCh38
rs75669887 Chr.7: 139341388-139341388 on Build GRCh38
rs771320141 Chr.7: 139341426-139341427 on Build GRCh38
rs766513651 Chr.7: 139341510-139341510 on Build GRCh38
rs759181790 Chr.7: 139341397-139341397 on Build GRCh38
rs750742734 Chr.7: 139341411-139341411 on Build GRCh38
rs776320891 Chr.7: 139341394-139341394 on Build GRCh38
rs764463286 Chr.7: 139341482-139341482 on Build GRCh38
rs752986208 Chr.7: 139341399-139341399 on Build GRCh38

View Full Product Details