Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52708356-52708581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC6
Entrez Gene ID
5706
Gene Name
proteasome 26S subunit, ATPase 6
Gene Aliases
CADP44, HEL-S-73, P44, SUG2, p42
UniGene
Hs.156171
Gene Chromosome Location
Chr.14: 52707163-52727998 on Build GRCh38
Transcripts
NM_002806.3 XM_017021472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149259 c.195G>C p.Q65H Chr.14: 52708376-52708376 on Build GRCh38
263145 c.153G>A p.Q51Q Chr.14: 52708376-52708376 on Build GRCh38
6757775 c.199G>T p.E67* Chr.14: 52708516-52708516 on Build GRCh38
6757774 c.241G>T p.E81* Chr.14: 52708516-52708516 on Build GRCh38
7149260 c.153G>C p.Q51H Chr.14: 52708376-52708376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921034715 Chr.14: 52708453-52708453 on Build GRCh38
rs370527009 Chr.14: 52708459-52708459 on Build GRCh38
rs759487433 Chr.14: 52708435-52708435 on Build GRCh38
rs755469022 Chr.14: 52708542-52708542 on Build GRCh38
rs910359972 Chr.14: 52708471-52708471 on Build GRCh38
rs772787080 Chr.14: 52708400-52708400 on Build GRCh38
rs377325167 Chr.14: 52708529-52708529 on Build GRCh38
rs1049815981 Chr.14: 52708407-52708407 on Build GRCh38
rs902887532 Chr.14: 52708539-52708540 on Build GRCh38
rs770428018 Chr.14: 52708417-52708417 on Build GRCh38
rs769435277 Chr.14: 52708387-52708387 on Build GRCh38
rs866188992 Chr.14: 52708398-52708398 on Build GRCh38
rs550393292 Chr.14: 52708408-52708408 on Build GRCh38
rs944564226 Chr.14: 52708474-52708474 on Build GRCh38
rs766447066 Chr.14: 52708511-52708511 on Build GRCh38
rs750607450 Chr.14: 52708463-52708463 on Build GRCh38
rs372995380 Chr.14: 52708447-52708447 on Build GRCh38
rs778124867 Chr.14: 52708413-52708413 on Build GRCh38
rs941211069 Chr.14: 52708431-52708431 on Build GRCh38
rs760478873 Chr.14: 52708457-52708457 on Build GRCh38
rs767478489 Chr.14: 52708444-52708444 on Build GRCh38
rs774187582 Chr.14: 52708433-52708433 on Build GRCh38
rs373347403 Chr.14: 52708477-52708477 on Build GRCh38
rs762971111 Chr.14: 52708494-52708494 on Build GRCh38

View Full Product Details